Skip to Content
Merck
All Photos(2)

Documents

AV44041

Sigma-Aldrich

Anti-SLC25A22 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-FLJ13044, Anti-GC1, Anti-Solute carrier family 25 (mitochondrial carrier: glutamate), member 22

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

36 kDa

species reactivity

human, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

Synthetic peptide directed towards the N terminal region of human SLC25A22

Application

Anti-SLC25A22 antibody produced in rabbit is suitable for western blotting at a concentration of 1.25μg/ml and for immunohistochemistry of paraffin-embedded tissue sections at a concentration of 4-8μg/ml.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunoprecipitation (1 paper)

Biochem/physiol Actions

SLC25A22 (GC1) is a mitochondrial glutamate carrier that transport glutamine across the inner mitochondrial membrane. The transport of glutamate to the cytoplasm from mitochondria by SLC25A22 regulates the secretion of insulin secretory vesicles in the islet β-cells.

Sequence

Synthetic peptide located within the following region: LQDAGRIAAQRKILAAQGQLSAQGGAQPSVEAPAAPRPTATQLTRDLLRS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sumiyo Morita et al.
PeerJ, 1, e162-e162 (2013-10-11)
Insulin secretion from pancreatic β-cells plays an essential role in blood glucose homeostasis and type 2 diabetes. Many genes are involved in the secretion of insulin and most of these genes can be targeted by microRNAs (miRNAs). However, the role
Giuseppe Fiermonte et al.
The Journal of biological chemistry, 277(22), 19289-19294 (2002-03-19)
The mitochondrial carriers are a family of transport proteins in the inner membranes of mitochondria. They shuttle substrates, metabolites, and cofactors through this membrane and connect cytoplasm functions with others in the matrix. Glutamate is co-transported with H(+) (or exchanged
Sudhir G Tattikota et al.
The Journal of biological chemistry, 290(33), 20284-20294 (2015-07-15)
In response to fasting or hyperglycemia, the pancreatic β-cell alters its output of secreted insulin; however, the pathways governing this adaptive response are not entirely established. Although the precise role of microRNAs (miRNAs) is also unclear, a recurring theme emphasizes
Sudhir G Tattikota et al.
Cell metabolism, 19(1), 122-134 (2013-12-24)
Pancreatic β cells adapt to compensate for increased metabolic demand during insulin resistance. Although the microRNA pathway has an essential role in β cell proliferation, the extent of its contribution is unclear. Here, we report that miR-184 is silenced in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service