Skip to Content
Merck
All Photos(2)

Documents

A7646

Sigma-Aldrich

Apyrase from potatoes

ATPase ≥60 units/mg protein, lyophilized powder (partially purified)

Synonym(s):

Adenosine 5′-diphosphatase, Adenosine 5′-triphosphatase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

potato

type

Grade III

form

lyophilized powder (partially purified)

ATPase activity

≥60 units/mg protein

mol wt

~49 kDa by gel filtration

composition

Protein, 30-60%

solubility

H2O: soluble 10 mg/mL, slightly hazy

application(s)

diagnostic assay manufacturing

foreign activity

Acid Phosphatase ≤5% of base activity

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Apyrase is used to hydrolyze nucleoside triphosphates and diphosphates. For hydrolysis of organic di and triphosphates, the optimal pH is 6, and for inorganic substrates, the optimal pH is 5.1 Apyrase, from Sigma, has been used in inhibition studies of platelet-aggregation. The enzyme has been used to assess the possibility of ATP involvement in intestinal lamina propria Th17 cell differentiation.
At least two isoenzymes are found in different varieties of S. tuberosum: one with a high ATPase/ADPase ratio (∼10) and another with a low ratio (∼1).
Reaction: ATP → ADP+Pi → AMP+2Pi.

Biochem/physiol Actions

Apyrase is found in all eukaryotes and some prokaryotes. Apyrase, from potato, has a crucial role in regulating growth and development. Apyrase is involved in the inactivation of synaptic ATP as a neurotransmitter following nerve stimulation and in the inhibition of ADP induced platelet aggregation to prevent thrombosis. Divalent metal ions are required for activity and best activity is observed with calcium ion at 5 mM. The molecular weight of the protein is found to be approximately 45 kDa.

Packaging

Sold on the basis of ATPase units

Other Notes

Mixture of high and low ratio isoenzymes.

Quality

Contains ≤ 5% acid phosphatase.

Unit Definition

One unit will liberate 1.0 μmole of inorganic phosphate from ATP or ADP per min at pH 6.5 at 30 °C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H J Kruse et al.
Arteriosclerosis and thrombosis : a journal of vascular biology, 14(11), 1821-1828 (1994-11-01)
The blocker of receptor-mediated calcium entry SK&F 96365 was used to evaluate the contribution of calcium influx to the formation of biologically active endothelial prostanoids and endothelium-derived relaxing factor (EDRF). SK&F 96365 inhibited histamine-stimulated calcium entry into human umbilical vein
Yen-Ta Chen et al.
Journal of tissue engineering and regenerative medicine, 13(12), 2155-2168 (2019-09-11)
This study tested the hypothesis that preactivated and disaggregated shape-changed platelet (PreD-SCP) therapy significantly protected rat kidney from ischemia-reperfusion (IR) injury. Adult-male Sprague-Dawley rats (n = 24) were equally categorized into Groups 1 (sham-operated control [SC]), 2 (SC + PreD-SCP)
Marko Gosak et al.
Life (Basel, Switzerland), 11(5) (2021-05-01)
Ca2+ homeostasis and signaling disturbances are associated with lens pathophysiology and are involved in cataract formation. Here, we explored the spatiotemporal changes in Ca2+ signaling in lens epithelial cells (LECs) upon local mechanical stimulation, to better understand the LECs' intercellular
Xiao-Jing Kuang et al.
Theranostics, 12(8), 3703-3718 (2022-06-07)
Rationale: Stress is a major risk factor for the development of depression. However, the underlying molecular mechanisms of stress vulnerability in depression are largely uncharacterized. Methods: P2X2 receptors (a major receptor for gliotransmitter-ATP) in the medial prefrontal cortex (mPFC) were
Flow-cytometric analysis of mouse platelet function.
Bernhard Nieswandt et al.
Methods in molecular biology (Clifton, N.J.), 272, 255-268 (2004-07-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service