Skip to Content
Merck
All Photos(2)

Documents

A6559

Sigma-Aldrich

Adenosine 5′-triphosphate disodium salt solution

Crystalline ATP, HPLC purified, aqueous solution for RNA transcription

Synonym(s):

ATP

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106305
eCl@ss:
32160414
PubChem Substance ID:
NACRES:
NA.52

Quality Level

form

liquid

concentration

100 mM in H2O (adjusted to pH 7 with Trizma®
base)

foreign activity

DNase, RNase, none detected

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[Na+].Nc1ncnc2n(cnc12)[C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O

InChI

1S/C10H16N5O13P3.2Na/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(26-10)1-25-30(21,22)28-31(23,24)27-29(18,19)20;;/h2-4,6-7,10,16-17H,1H2,(H,21,22)(H,23,24)(H2,11,12,13)(H2,18,19,20);;/q;2*+1/p-2/t4-,6-,7-,10-;;/m1../s1

InChI key

TTWYZDPBDWHJOR-IDIVVRGQSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Adenosine 5′-triphosphate disodium salt solution is suitable for use in molecular biology applications such as kinase reactions or DNA-dependent RNA polymerase transcription.

Application

Adenosine 5′-triphosphate disodium salt solution has been used:
  • as a standard to evaluate the extracellular ATP release from marginal cells of stria vascular
  • as a component of the internal pipette solution to establish a recording electrode to study neurite stretching
  • in in vitro VPS34 (catalytic subunit of the phosphatidylinositol 3-kinase (PtdIns3K) complex) activity assay
  • as a component of lysis and resealing buffers for ATP studies
  • in spectrophotometric measurement of H+ transport in fluorescein isothiocyanate (FITC)-dextran loaded A549 (adenocarcinomic human alveolar basal epithelial cells) and HeLa cells
  • to induce pyroptosis in lipopolysaccharide treated fibroblast-like synoviocytes (FLSs)

Biochem/physiol Actions

Adenosine 5′-triphosphate (ATP) is an important energy currency in all living organisms. It possesses high phosphate transfer potential. ATP is involved in several biological processes such as membrane transport, muscle contraction and synthesis, and degradation of biological molecules. It plays a role as an intracellular and extracellular signaling molecule in certain cellular processes such as cell motility, organ development, neurotransmission, and insulin secretion.

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yahui Huang et al.
Archiv der Pharmazie (2018-06-29)
Inhibition of spleen tyrosine kinase (Syk) is a promising strategy for the treatment of various allergic and autoimmune disorders such as asthma, rheumatoid arthritis, and allergic rhinitis. Previously, a Syk inhibitor with novel indazole scaffold was discovered by structure-based virtual
James Franklin Tucker et al.
PLoS genetics, 12(2), e1005873-e1005873 (2016-02-20)
Epigenetic gene silencing plays a critical role in regulating gene expression and contributes to organismal development and cell fate acquisition in eukaryotes. In fission yeast, Schizosaccharomyces pombe, heterochromatin-associated gene silencing is known to be mediated by RNA processing pathways including
Cooper GM
The Cell: A Molecular Approach (2000)
Identification of small molecule inhibitors of phosphatidylinositol 3-kinase and autophagy.
Farkas T
The Journal of Biological Chemistry, 286(45), 38904-38912 (2011)
Shaojian Chen et al.
Cell cycle (Georgetown, Tex.), 20(12), 1107-1121 (2021-06-08)
Studies have found that cell pyroptosis is involved in the occurrence and development of rheumatoid arthritis (RA). Hsa_circ_0044235 has been found to be significantly low-expressed in RA patients. The purpose of this research was to reveal the regulatory mechanism of

Articles

Understand how mRNA vaccines induce immunity. and read how synthetic mRNA is prepared for vaccine immunogens and other biopharmaceuticals. Find reagents for synthesis of mRNA.

Understand how mRNA vaccines induce immunity. and read how synthetic mRNA is prepared for vaccine immunogens and other biopharmaceuticals. Find reagents for synthesis of mRNA.

Understand how mRNA vaccines induce immunity. and read how synthetic mRNA is prepared for vaccine immunogens and other biopharmaceuticals. Find reagents for synthesis of mRNA.

Understand how mRNA vaccines induce immunity. and read how synthetic mRNA is prepared for vaccine immunogens and other biopharmaceuticals. Find reagents for synthesis of mRNA.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service