Skip to Content
Merck
All Photos(1)

Key Documents

A7430

Sigma-Aldrich

Azadirachtin

~95%

Synonym(s):

azadirachtin A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H44O16
CAS Number:
Molecular Weight:
720.71
MDL number:
UNSPSC Code:
12352212
PubChem Substance ID:
NACRES:
NA.28

Assay

~95%

Quality Level

form

solid

application(s)

agriculture
environmental

storage temp.

−20°C

SMILES string

[H]C1([H])C2OC3([H])OC=CC3(O)C1C4(C)OC24[C@@]5(C)[C@@H](O)[C@@H]6OC[C@]7([C@H](C[C@H](OC(=O)C(C)=CC)[C@]8(COC(O)([C@H]58)C(=O)OC)[C@@]67[H])OC(C)=O)C(=O)OC

InChI

1S/C35H44O16/c1-8-15(2)24(38)49-18-12-19(48-16(3)36)32(26(39)43-6)13-46-21-22(32)31(18)14-47-34(42,27(40)44-7)25(31)29(4,23(21)37)35-20-11-17(30(35,5)51-35)33(41)9-10-45-28(33)50-20/h8-10,17-23,25,28,37,41-42H,11-14H2,1-7H3/t17?,18-,19?,20?,21+,22+,23?,25?,28?,29?,30?,31?,32-,33?,34-,35?/m0/s1

InChI key

FTNJWQUOZFUQQJ-CFYAVYSVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Azadirachtin has been used as a standard for high performance liquid chromatography (HPLC) and for quantification in suspension cultures of Azadirachta indica (A. Juss).

Biochem/physiol Actions

Triterpenoid found in need tree seeds, azadiractin suppresses feeding by many insect species and disrupts growth of most insect and other arthropod species, while having very low mammalian toxicity. Promising as a natural pesticide.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

related product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Benshui Shu et al.
Scientific reports, 7(1), 13231-13231 (2017-10-19)
As an important botanical pesticide, azadirachtin demonstrates broad insecticidal activity against many agricultural pests. The results of a previous study indicated the toxicity and apoptosis induction of azadirachtin in Spodoptera frugiperda Sf9 cells. However, the lack of genomic data has
Statistical media optimization for cell growth and azadirachtin production in Azadirachta indica (A. Juss) suspension cultures
Prakash G and Srivastava AK
Process Biochemistry (Oxford, United Kingdom), 40(12), 3795-3800 (2005)
Cecilia Stahl Vieira et al.
Developmental and comparative immunology, 114, 103864-103864 (2020-09-13)
Rhodnius prolixus is an insect vector of two flagellate parasites, Trypanosoma rangeli and Trypanosoma cruzi, the latter being the causative agent of Chagas disease in Latin America. The R. prolixus neuroendocrine system regulates the synthesis of the steroid hormone ecdysone
Torranis Ruttanaphan et al.
Insects, 11(10) (2020-10-15)
Spodoptera frugiperda, a highly polyphagous insect pest from America, has recently invaded and widely spread throughout Africa and Asia. Effective and environmentally safe tools are needed for successful pest management of this invasive species. Natural molecules extracted from plants offer
Pragya Srivastava et al.
Carcinogenesis, 33(11), 2199-2207 (2012-08-24)
Azadirachta indica, commonly known as neem, has a wide range of medicinal properties. Neem extracts and its purified products have been examined for induction of apoptosis in multiple cancer cell types; however, its underlying mechanisms remain undefined. We show that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service