Skip to Content
Merck
All Photos(2)

Key Documents

A4910

Sigma-Aldrich

DL-2-Amino-3-phosphonopropionic acid

Synonym(s):

3-Phosphono-DL-alanine, AP-3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H8NO5P
CAS Number:
Molecular Weight:
169.07
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.32

form

powder

mp

227-229 °C (lit.)

SMILES string

NC(CP(O)(O)=O)C(O)=O

InChI

1S/C3H8NO5P/c4-2(3(5)6)1-10(7,8)9/h2H,1,4H2,(H,5,6)(H2,7,8,9)

InChI key

LBTABPSJONFLPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Potent metabotropic glutamate receptor antagonist.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Banno et al.
The Journal of comparative neurology, 402(2), 252-263 (1998-12-09)
The intraventricular administrations of L-glutamate or trans-1-amino-1,3-cyclopentanedicarboxylic acid (t-ACPD), which is an agonist for the metabotropic glutamate receptor, induced conformational changes of the smooth endoplasmic reticulum (SER) to form lamellar bodies, consisting of stacks of flattened cisterns in Purkinje cell
Tine Pokorn et al.
PloS one, 12(9), e0184528-e0184528 (2017-09-09)
Viroids, the smallest known pathogens, unable to encode any proteins, can cause severe diseases in their host plants. One of the proposed mechanisms of their pathogenicity includes silencing the host's genes via viroid-derived small RNAs, which are products of the
Tao Wang et al.
Molecular cancer research : MCR, 17(2), 398-408 (2018-09-30)
The αvβ3 integrin is involved in various physiologic and pathologic processes such as wound healing, angiogenesis, tumor growth, and metastasis. The impact of αvβ3 integrin on the radiosensitivity of prostate cancer cells and the molecular mechanism controlling cell survival in
Akari Takai et al.
International journal of molecular sciences, 21(17) (2020-09-10)
Developmental and epileptic encephalopathies (DEEs) are the spectrum of severe epilepsies characterized by early-onset, refractory seizures occurring in the context of developmental regression or plateauing. Early infantile epileptic encephalopathy (EIEE) is one of the earliest forms of DEE, manifesting as
G Pellegri et al.
The European journal of neuroscience, 10(1), 272-280 (1998-09-30)
In view of the neurotrophic effect of vasoactive intestinal peptide (VIP), the regulation of brain-derived neurotrophic factor (BDNF) expression by VIP and the related peptide pituitary adenylate cyclase-activating polypeptide (PACAP) was analysed by Northern blot in primary cultures of cortical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service