Skip to Content
Merck
All Photos(1)

Key Documents

88185

Sigma-Aldrich

Tetrazole solution

Tetrazole solution

suitable for DNA synthesis, filtered through a 1 μm filter, ~0.45 M in acetonitrile

Synonym(s):

1H-Tetrazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH2N4
CAS Number:
Molecular Weight:
70.05
Beilstein:
105799
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
technique(s):
DNA synthesis: suitable
bp:
84 °C (lit.)
Pricing and availability is not currently available.

form

liquid

Quality Level

quality

filtered through a 1 μm filter

concentration

~0.45 M in acetonitrile

technique(s)

DNA synthesis: suitable

impurities

≤0.003% water

bp

84 °C (lit.)

mp

156-158 °C (lit.)

density

0.798 g/mL at 20 °C

SMILES string

c1nnn[nH]1

InChI

1S/CH2N4/c1-2-4-5-3-1/h1H,(H,2,3,4,5)

InChI key

KJUGUADJHNHALS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The product is ~0.45M solution of tetrazole in acetonitrile. Tetrazole ring plays significant role in various synthetic and industrial processes.[1]

Application

Tetrazole solution is useful for DNA synthesis. It may be employed as a catalyst for the in situ synthesis of deoxyribonucleoside phosphoramidites.[2] It may be used in the preparation of following anionic nucleotide-lipids:[3]
  • thymidine 3′-(1,2-dilauroyl-sn-glycero-3-phosphate) , diC12-3′-dT
  • thymidine 3′-(1,2-dimyristoyl-sn-glycero-3-phosphate) diC14- 3′-dT
  • thymidine-3′-(1,2-dipalmitoyl-sn-glycero-3-phosphate) diC16- 3′-dT

Caution

Saturated solution at room temperature; storage below room temperature causes precipitation.

Other Notes

Catalyst used in the phosphite triester method of oligonucleotide synthesis for the coupling of (dialkylamino)phosphines[4]

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

41.0 °F - closed cup

Flash Point(C)

5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Azidoazomethine-tetrazole isomerism in solution: A thermochemical study.
Cubero E, et al.
The Journal of Organic Chemistry, 63(7), 2354-2356 (1998)
A D Barone et al.
Nucleic acids research, 12(10), 4051-4061 (1984-05-25)
Deoxynucleoside phosphoramidites can be prepared in good yield from deoxynucleosides, bis- dialkylaminophosphines , and the corresponding dialkylamine hydrotetrazolide or tetrazole as catalysts. These phosphoramidites generated in situ lead to the direct synthesis of deoxyoligonucleotides on polymer supports.
Salim Khiati et al.
Bioconjugate chemistry, 20(9), 1765-1772 (2009-08-29)
A family of new anionic nucleotide based lipids featuring thymidine-3'-monophosphate as nucleotide and 1,2-diacyl-sn-glycerol as lipid moiety for in vitro delivery of nucleic acids is described. The nucleotide lipids were prepared in three steps starting from 1,2-diacyl-sn-glycerols and 2'-deoxythymidine-3'-phosphoramidite. Gel
J Nielsen et al.
Nucleic acids research, 14(18), 7391-7403 (1986-09-25)
Deoxyribonucleoside phosphoramidites are prepared in situ from 5'-O,N-protected deoxyribonucleosides and 2-cyanoethyl N,N,N',N'-tetraisopropylphosphorodiamidite with tetrazole as catalyst, and the solutions applied directly on an automatic solid-phase DNA synthesizer. Using LCAA-CPG support and a cycle time of 12.5 min, oligonucleotides of 16-25
Zhipeng Yu et al.
Journal of the American Chemical Society, 133(31), 11912-11915 (2011-07-09)
Photoactivatable fluorescent probes are invaluable tools for the study of biological processes with high resolution in space and time. Numerous strategies have been developed in generating photoactivatable fluorescent probes, most of which rely on the photo-"uncaging" and photoisomerization reactions. To

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service