Skip to Content
Merck
All Photos(3)

Key Documents

S2255

Sigma-Aldrich

Safranin O

Dye content ≥85 %, Powder

Synonym(s):

Basic Red 2, Cotton Red, Gossypimine, Safranin T, Safranin Y or A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19ClN4
CAS Number:
Molecular Weight:
350.84
Colour Index Number:
50240
Beilstein:
3924099
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Safranin O, Dye content ≥85 %

form

powder

Quality Level

composition

Dye content, ≥85%

technique(s)

microbe id | staining: suitable

pH

10 (20 °C, 10 g/L)

solubility

water: 1 mg/mL, clear, dark red to very dark red and red purple

εmax

1250-1650 at 530-534 nm in 50% ethanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].Cc1cc2nc3cc(C)c(N)cc3[n+](-c4ccccc4)c2cc1N

InChI

1S/C20H18N4.ClH/c1-12-8-17-19(10-15(12)21)24(14-6-4-3-5-7-14)20-11-16(22)13(2)9-18(20)23-17;/h3-11H,1-2H3,(H3,21,22);1H

InChI key

OARRHUQTFTUEOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Safranin O is a cationic dye, which displays less significant metachromasia. It is used to detect any variations in articular disease. Safranin O binds specifically to polyanions. It detects the amount of proteoglycan present in cartilages. Safranin O is used as a marker for glycosaminoglycan depletion in osteoarthritis. Safranin is used as a counter stain in Gram staining.

Application

Safranin O has been used in the histological analysis of femurs. It has also been used to detect the presence of glycosaminoglycans in the inner body of meniscus structures.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Porous CaP/silk composite scaffolds to repair femur defects in an osteoporotic model
Cheng N, et al.
Journal of Materials Science. Materials in Medicine, 24(8), 1963-1975 (2013)
Yanting Gu et al.
Polymers, 11(4) (2019-04-18)
Solid acids have been proposed as a hydrolytic agent for wood biomass dissolution. In this work, we presented an environmentally friendly physicochemical treatment to leave behind cellulose, dissolve hemicellulose, and remove lignin from poplar wood. Several pretreatments, such as autohydrolysis
Variability in particle degradation of four commonly employed dental bone grafts
Yang S, et al.
Clinical Implant Dentistry and Related Research, 17(5), 996-1003 (2015)
Delivery of PDGF-B and BMP-7 by mesoporous bioglass/silk fibrin scaffolds for the repair of osteoporotic defects
Zhang Y, et al.
Biomaterials, 33(28), 6698-6708 (2012)
Delivery of PDGF-B and BMP-7 by mesoporous bioglass/silk fibrin scaffolds for the repair of osteoporotic defects
Yufeng Zhang
Biomaterials (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service