Skip to Content
Merck
All Photos(1)

Key Documents

75999

Sigma-Aldrich

Atto 680 NHS ester

BioReagent, suitable for fluorescence, ≥90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Assay

≥90% (HPLC)
≥90% (coupling of degree)

form

powder

manufacturer/tradename

ATTO-TEC GmbH

λ

in methanol (with trifluoroacetic acid)

UV absorption

λ: 667-673 nm Amax

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

[S](=O)(=O)([O-])CC1=CC(N(c3c1cc4nc5c([o+]c4c3)cc6c(c5)CCCN6CC)CCCC(=O)ON2C(=O)CCC2=O)(C)C

InChI

1S/C31H34N4O8S/c1-4-33-11-5-7-19-13-22-26(15-24(19)33)42-27-16-25-21(14-23(27)32-22)20(18-44(39,40)41)17-31(2,3)34(25)12-6-8-30(38)43-35-28(36)9-10-29(35)37/h13-17H,4-12,18H2,1-3H3

InChI key

PVQYIKUDSQLKDA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Molar absorption 120,000 1/M cm, abs: 675 nm, em: 705 nm, QY=0.3, Tfl 1.7 ns (unpublished data); excitation energy acceptor in an acceptor/donor zeolite system.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Energy transfer from dye-zeolite L antenna crystals to bulk silicon.
Stefan Huber et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 5(2), 239-242 (2004-03-25)
Irene Drude et al.
Biochemical and biophysical research communications, 363(1), 24-29 (2007-09-11)
Over the past two decades, the structure and mechanism of catalytic RNA have been extensively studied; now ribozymes are understood well enough to turn them into useful tools. After we have demonstrated the twin ribozyme mediated insertion of additional nucleotides
Slawomir Gwiazda et al.
Biochimie, 94(7), 1457-1463 (2012-04-03)
The RNA-world-theory is one possible explanation of how life on earth has evolved. In this context it is of high interest to search for molecular systems, capable of self-organization into structures with increasing complexity. We have engineered a simple catalytic
Philip Tinnefeld et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 6(2), 217-222 (2005-03-09)
Light-harvesting complexes, one of nature's supreme examples of nanoscale engineering, have inspired researchers to construct molecular optical devices, such as photonic wires, which are optimised for efficient transfer of excited-state energy over large distances. The control parameters for the design
The design of a peptide linker group to enhance the SERS signal intensity of an atto680 dye-nanoparticle system.
Drake, P., et al.
Journal of Raman Spectroscopy, 41, 1248-1253 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service