Skip to Content

79636

Atto 590 NHS ester

BioReagent, suitable for fluorescence, ≥90.0% (degree of coupling)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C41H42ClN3O11
CAS Number:
Molecular Weight:
788.24
UNSPSC Code:
12352108
NACRES:
NA.32
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

Quality Segment

assay

≥90.0% (HPLC), ≥90.0% (degree of coupling)

form

powder

manufacturer/tradename

ATTO-TEC GmbH

λ

in ethanol (with 0,1% trifluoroacetic acid)

UV absorption

λ: 597-663 nm Amax

suitability

suitable for fluorescence

detection method

fluorometric

storage temp.

−20°C

SMILES string

[Cl](=O)(=O)(=O)[O-].[N+]1(=C2C(=CC3=C(c6c(cc7c(c6)C(=CC(N7CC)(C)C)C)OC3=C2)c4c(ccc(c4)C(=O)ON5C(=O)CCC5=O)C(=O)O)C(=CC1(C)C)C)CC

InChI

1S/C41H41N3O7.ClHO4/c1-9-42-31-18-33-29(16-26(31)22(3)20-40(42,5)6)37(30-17-27-23(4)21-41(7,8)43(10-2)32(27)19-34(30)50-33)28-15-24(11-12-25(28)38(47)48)39(49)51-44-35(45)13-14-36(44)46;2-1(3,4)5/h11-12,15-21H,9-10,13-14H2,1-8H3;(H,2,3,4,5)

InChI key

SHJDVERDESTYRQ-UHFFFAOYSA-N

General description

Atto 590 is a novel fluorescent label belonging to the class of Rhodamine dyes. The dye is designed for application in the area of life science, e.g. labeling of DNA, RNA or proteins. The Atto dyes are a series of fluorescent dyes that provide all the crucial properties required for modern fluorescent technologies, such as fluorescence microscopy, flow-cytometry, fluorescence in situ hybridization (FISH), receptor binding assays or enzyme assays. The dye is highly suitable for single-molecule detection applications and high-resolution microscopy. The NHS-esters are used in common conjugation protocols.

Application

Atto 590 NHS ester may be suitable for use in site-specific labeling of human embryonic kidney (HEK293T) cell lysates for western blotting, fluorescence, and widefield microscopy studies.
Molar absorption 120,000 1/M cm, abs: 593 nm, em: 620 nm, QY=0.93, Tfl 4.0 ns (unpublished data)

Features and Benefits

  • Absorption maxima cover a range from 590 to 620 nm.
  • Structural properties support extraordinary photostability.
  • High fluorescence quantum yield and high thermal stability.
  • The fluorescence characteristics of the dye are quite insensitive to environmental changes (e.g., pH changes). This holds for both excitation and emission wavelengths and for the decay time of their fluorescence emission.
  • Compatible with common labeling procedures for proteins and amino-functionalized oligonucleotides.

Other Notes

Study of multistep energy transfer in a single photonic wire by FRET; employed in fluorescence energy transfer, FRET, scanning microscopy.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library