Skip to Content
Merck
All Photos(1)

Key Documents

CDS020796

Sigma-Aldrich

Glycyrrhizin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H62O16
Molecular Weight:
822.93
UNSPSC Code:
12352200
PubChem Substance ID:

description

AldrichCPR

form

solid

SMILES string

[H][C@@]12C[C@](CC[C@@]1(CC[C@@]3(C2=CC([C@@]4([C@]5(CC[C@@H](C(C)([C@@]5(CC[C@@]34C)[H])C)O[C@H]6O[C@H](C(O)=O)[C@H]([C@@H]([C@H]6O[C@@H]7O[C@H](C(O)=O)[C@H]([C@@H]([C@H]7O)O)O)O)O)C)[H])=O)C)C)(C(O)=O)C

InChI

1S/C42H62O16/c1-37(2)21-8-11-42(7)31(20(43)16-18-19-17-39(4,36(53)54)13-12-38(19,3)14-15-41(18,42)6)40(21,5)10-9-22(37)55-35-30(26(47)25(46)29(57-35)33(51)52)58-34-27(48)23(44)24(45)28(56-34)32(49)50/h16,19,21-31,34-35,44-48H,8-15,17H2,1-7H3,(H,49,50)(H,51,52)(H,53,54)/t19-,21-,22-,23-,24-,25-,26-,27+,28-,29-,30+,31+,34-,35-,38+,39-,40-,41+,42+/m0/s1

InChI key

LPLVUJXQOOQHMX-QWBHMCJMSA-N

General description

Glycyrrhizin is a plant glycoside derived from the liquorice (Glycyrrhiza glabra) plant root. It is a sweet-tasting molecule sweeter than sucrose (about 30-50 times), but it does not influence the glycemic index since it is not metabolized. It is often used as a natural emulsifier and gel-forming agent in foodstuffs,, beverages, and cosmetics.

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flavors
Introduction to the Chemistry of Food (2020)
Glycyrrhizin: An alternative drug for the treatment of COVID-19 infection and the associated respiratory syndrome?
Christian B and Gerard V
Pharmacology & Therapeutics, 214, 107618-107618 (2020)
Carina Dargel et al.
Biophysical journal, 118(10), 2411-2425 (2020-04-26)
In this study, we investigated the conversion of lipid bicelles into vesicles in the case of a system composed of the phospholipid 1,2-dimyristoyl-sn-glycero-3-phosphocholine (DMPC) and the saponin glycyrrhizin in the presence of sucrose. Glycyrrhizin is a biosurfactant present in the
Christian Bailly et al.
Pharmacology & therapeutics, 214, 107618-107618 (2020-06-28)
Safe and efficient drugs to combat the current COVID-19 pandemic are urgently needed. In this context, we have analyzed the anti-coronavirus potential of the natural product glycyrrhizic acid (GLR), a drug used to treat liver diseases (including viral hepatitis) and
Noha M Shafik et al.
Archives of biochemistry and biophysics, 675, 108095-108095 (2019-09-03)
Rheumatoid arthritis (RA) is a systemic chronic autoimmune disease manifested by joint destruction and deformity, hence decreasing patient's life quality. The aim of the present work is to explore the mechanistic effects of glycyrrhizin (GL)and/or platelet rich plasma (PRP) treatment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service