Skip to Content
Merck
All Photos(1)

Key Documents

63559

Sigma-Aldrich

D-Mannitol

BioUltra, ≥99.0% (sum of enantiomers, HPLC)

Synonym(s):

Mannite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
Beilstein:
1721898
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

product line

BioUltra

Quality Level

Assay

≥99.0% (sum of enantiomers, HPLC)

form

powder

optical activity

[α]20/D +24.0±1° in borax solution acc. to Ph Eur

impurities

insoluble matter, passes filter test
reducing sugars, complies
≤0.0072% free acid (as CH3COOH)

ign. residue

≤0.01% (as SO4)

loss

≤0.05% loss on drying, 20 °C (HV)

color

white

pH

5.0-6.5 (25 °C, 1 M in H2O)

mp

166-168 °C
167-170 °C (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.04
λ: 280 nm Amax: 0.04

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4-,5-,6-/m1/s1

InChI key

FBPFZTCFMRRESA-KVTDHHQDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

D-Mannitol is used as an osmolyte and transport facilitator in a variety of biological in vivo and in vitro applications, such as improvement of blood-brain barrier transport and hyperosmolar therapy. D-Mannitol is used as a substrate to identify, differentiate and characterized enzymes such as mannitol dehydrogenase(s). D-Mannitol is used in identify mannitol metabolizing bacteria.

Biochem/physiol Actions

A sugar alcohol sweet tastant. Used in sweetness inhibition studies.

Other Notes

To gain a comprehensive understanding of our extensive range of Sugar alcohols for your research, we encourage you to visit our Carbohydrates Category page.
Used in discontinuous gradient centrifugation for purification of plant protoplasts. Plasmolyticum that reversibly inhibits phenylalanine ammonia-lyase activity of protoplasts at concentrations exceeding 50 mM. Component of the liquid stationary phase in the GLC separation of nitroxylene and xylenol isomers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

B. Fritig
Phytochemistry, 16, 895-895 (1977)
Anna Stepanova et al.
Antioxidants & redox signaling, 31(9), 608-622 (2019-05-01)
Aims: Brain ischemia/reperfusion (I/R) is associated with impairment of mitochondrial function. However, the mechanisms of mitochondrial failure are not
B.G. Hughes
Biochem. Physiol. Pflanz., 172, 223-223 (1978)
Separation behaviour of some isomeric organic compounds on sugars, sugar alcohols and their mixed phases by gas-liquid chromatography.
Ono, A.
Journal of Chromatography A, 197, 251-251 (1980)
Heikki O Koskela et al.
Chest, 125(6), 1985-1992 (2004-06-11)
To define whether coughing during mannitol challenge is a nonspecific side effect of this challenge or is associated with asthma. A prospective study. University hospital. Thirty-seven steroid-naive, asthmatic subjects and 10 healthy subjects. The participants completed a symptom questionnaire, recorded

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service