Skip to Content
Merck
All Photos(3)

Key Documents

M1902

Sigma-Aldrich

D-Mannitol

≥98% (GC), suitable for plant cell culture

Synonym(s):

Mannite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
Beilstein:
1721898
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21

Assay

≥98% (GC)

form

powder

technique(s)

cell culture | plant: suitable

color

white

useful pH range

5.0-6.5 (25 °C, 182 g/L)

mp

167-170 °C (lit.)

solubility

water: soluble 182 g/L at 20 °C (68 °F)

application(s)

agriculture

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4-,5-,6-/m1/s1

InChI key

FBPFZTCFMRRESA-KVTDHHQDSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Effect of different proportions of glycerol and D-mannitol as plasticizer on the properties of extruded corn starch.: This paper examines how varying proportions of glycerol and D-Mannitol affect the mechanical properties of extruded corn starch, which is critical for the development of biodegradable packaging materials (Xu et al., 2023).

  • Exposure of Helicobacter pylori to clarithromycin in vitro resulting in the development of resistance and triggers metabolic reprogramming associated with virulence and pathogenicity.: The study explores how D-Mannitol contributes to metabolic reprogramming in Helicobacter pylori when exposed to clarithromycin, offering insights into antibiotic resistance mechanisms and potential therapeutic targets (Rosli et al., 2024).

Biochem/physiol Actions

A sugar alcohol sweet tastant. Used in sweetness inhibition studies.

Other Notes

To gain a comprehensive understanding of our extensive range of Sugar alcohols for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Heikki O Koskela et al.
Chest, 125(6), 1985-1992 (2004-06-11)
To define whether coughing during mannitol challenge is a nonspecific side effect of this challenge or is associated with asthma. A prospective study. University hospital. Thirty-seven steroid-naive, asthmatic subjects and 10 healthy subjects. The participants completed a symptom questionnaire, recorded
Heikki O Koskela et al.
Chest, 124(4), 1341-1349 (2003-10-14)
To compare the sensitivity and validity of mannitol, histamine, and cold air challenges to demonstrate the effect of inhaled corticosteroids (ICS) in asthma. A prospective study. Seventeen patients with recently diagnosed, steroid-naive asthma who fulfilled the diagnostic criteria of Finnish
Cecilio F Caldeira et al.
Nature communications, 5, 5365-5365 (2014-11-06)
Circadian rhythms enable plants to anticipate daily environmental variations, resulting in growth oscillations under continuous light. Because plants daily transpire up to 200% of their water content, their water status oscillates from favourable during the night to unfavourable during the
Pär Matsson et al.
Journal of medicinal chemistry, 48(2), 604-613 (2005-01-22)
The influence of different drug transport routes in intestinal drug permeability screening assays was studied. Three experimental models were compared: the small-intestine-like 2/4/A1 cell model, which has a leaky paracellular pathway, the Caco-2 cell model, which has a tighter paracellular
Diana Bilton et al.
Thorax, 69(12), 1073-1079 (2014-09-24)
Bronchiectasis is characterised by excessive production of mucus and pulmonary exacerbations. Inhaled osmotic agents may enhance mucociliary clearance, but few long-term clinical trials have been conducted. To determine the impact of inhaled mannitol on exacerbation rates in patients with non-cystic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service