Skip to Content
Merck
All Photos(1)

Key Documents

62661

Sigma-Aldrich

LR white resin

standard

Synonym(s):

Acryl resin LR white

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.21
Pricing and availability is not currently available.

impurities

bisphenol A dimethyl acrylate resin

color

colorless to almost colorless

density

1.0 g/cm3

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Benzoyl peroxide (B5907) is specifically formulated as an initiator for these resins.
LR White Resin is a very low viscosity resin, non-toxic and suitable for various applications in both light and Electron Microscopy. Major advantage of LR White sections is that they show minimal non-specific staining. Sections of polymerized resin are hydrophilic, thus immunocytochemistry reagents easily penetrate into the section. Its low viscosity makes LR White an ideal tool for infiltrating plant tissues as well as decalcified bone and teeth.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carola Pritzkow et al.
Tree physiology, 40(2), 215-229 (2019-12-21)
Understanding which hydraulic traits are under genetic control and/or are phenotypically plastic is essential in understanding how tree species will respond to rapid shifts in climate. We quantified hydraulic traits in Eucalyptus obliqua L'Her. across a precipitation gradient in the
A Matsuno et al.
The Histochemical journal, 30(2), 105-109 (1999-04-07)
An improved new method for the simultaneous visualization of mRNA and encoded protein in LR White resin-embedded specimens is described. This pre-embedding electron microscopical in situ hybridization (procedure) localized rat growth hormone mRNA specifically as high electron-density products on the
Xin Zhu et al.
Development (Cambridge, England), 147(1) (2019-12-13)
Cilia rotation-driven nodal flow is crucial for the left-right (L-R) break in symmetry in most vertebrates. However, the mechanism by which the flow signal is translated to asymmetric gene expression has been insufficiently addressed. Here, we show that Hedgehog (Hh)
Gina Puska et al.
Neurobiology of disease, 114, 140-152 (2018-03-06)
Intracellular deposition of pathologically altered α-synuclein mostly in neurons characterises Parkinson's disease (PD), while its accumulation predominantly in oligodendrocytes is a feature of multiple system atrophy (MSA). Recently a prion-like spreading of pathologic α-synuclein has been suggested to play a
S H Brorson
Micron (Oxford, England : 1993), 29(6), 439-443 (1999-03-11)
The purpose of this study was to examine the intensity of the immunogold labeling of kappa light chains as single molecules and as parts of whole immunoglobulin molecules in LR-White sections and epoxy sections both practically and theoretically. Human renal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service