Skip to Content
Merck
All Photos(1)

Key Documents

62476

Sigma-Aldrich

Lithium chloride

BioUltra, for molecular biology, anhydrous, ≥99.0% (AT)

Synonym(s):

LiCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiCl
CAS Number:
Molecular Weight:
42.39
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.26
Assay:
≥99.0% (AT)
grade:
for molecular biology
form:
powder or crystals
solubility:
H2O: 1 M at 20 °C, clear, colorless

grade

for molecular biology

Quality Level

product line

BioUltra

Assay

≥99.0% (AT)

form

powder or crystals

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected
≤0.001% total nitrogen (N)

loss

≤1% loss on drying, 110 °C

pH

5.0-7.5 (25 °C, 1 M in H2O)

mp

605 °C (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.2 mg/kg
Ba: ≤10 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤10 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.01
λ: 280 nm Amax: 0.01

SMILES string

[Li+].[Cl-]

InChI

1S/ClH.Li/h1H;/q;+1/p-1

InChI key

KWGKDLIKAYFUFQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • The preconditioning of lithium promotes mesenchymal stem cell-based therapy for the degenerated intervertebral disc via upregulating cellular ROS: This study explores the use of lithium chloride in enhancing the efficacy of mesenchymal stem cells for intervertebral disc regeneration. Lithium chloride′s role in modulating cellular responses and oxidative stress mechanisms presents a potential application in developing advanced IVD (In Vitro Diagnostics) products that rely on cellular technologies (Zhu et al., 2021).


Other Notes

Precipitation of RNA; Transformation of intact yeast cells treated with alkali cations

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Cathala et al.
DNA (Mary Ann Liebert, Inc.), 2(4), 329-335 (1983-01-01)
A method for isolation of large, translationally active RNA species is presented. The procedure involves homogenization of cells or tissues in 5 M guanidine monothiocyanate followed by direct precipitation of RNA from the guanidinium by 4 M LiCl. Modifications are
D.M. Wallace
Methods in Enzymology, 152, 48-48 (1987)
C Auffray et al.
European journal of biochemistry, 107(2), 303-314 (1980-06-01)
A procedure is described for the large-scale purification of light (L) and heavy (H) chain mRNAs from plasmacytomas produced in mice. Intact RNA is selectively precipitated in high yield from frozen tumors homogenized in 3 M LiCl and 6 M
H Ito et al.
Journal of bacteriology, 153(1), 163-168 (1983-01-01)
Intact yeast cells treated with alkali cations took up plasmid DNA. Li+, Cs+, Rb+, K+, and Na+ were effective in inducing competence. Conditions for the transformation of Saccharomyces cerevisiae D13-1A with plasmid YRp7 were studied in detail with CsCl. The
Pengyi Yang et al.
Cell systems, 8(5), 427-445 (2019-05-13)
Pluripotency is highly dynamic and progresses through a continuum of pluripotent stem cell states. The two states that bookend the pluripotency continuum, naive and primed, are well characterized, but our understanding of the intermediate states and transitions between them remains

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service