Skip to Content
Merck
All Photos(1)

Key Documents

73036

Sigma-Aldrich

Lithium chloride

puriss. p.a., anhydrous, ≥99.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiCl
CAS Number:
Molecular Weight:
42.39
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
form:
powder or crystals

vapor pressure

1.33 hPa ( 547 °C)

grade

puriss. p.a.

Assay

≥99.0% (AT)

form

powder or crystals

loss

<1% loss on drying, 110°C

pH

5.5-7.5 (25 °C, 50 mg/mL in H2O)

mp

605 °C (lit.)

anion traces

sulfate (SO42-): ≤50 mg/kg

cation traces

Ba: ≤10 mg/kg
Ca: ≤20 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤10 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[Li+].[Cl-]

InChI

1S/ClH.Li/h1H;/q;+1/p-1

InChI key

KWGKDLIKAYFUFQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Lithium chloride (LiCl) is a lithium salt. It participates in the Horner-Wadsworth-Emmons reactions of base-sensitive aldehydes and phosphonates. LiCl/N,N-Dimethylacetamide (DMAc) solvent system has been employed for the dissolution of cellulose and synthesis of various cellulose derivatives (cellulose esters, carbamates, sulfonates and ethers). It can be synthesized from lithium carbonate (Li2CO3).

Application

Lithium chloride (LiCl) may be used in the synthesis of lithium perchlorate by reacting with perchloric acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Horner-wadsworth-emmons reaction: Use of lithium chloride and an amine for base-sensitive compounds.
Blanchette MA, et al.
Tetrahedron Letters, 25(21), 2183-2186 (1984)
Derivatization of cellulose in lithium chloride and N-N-dimethylacetamide solutions.
McCormick CL and Callais PA.
Polymer, 28(13), 2317-2323 (1987)
Solution studies of cellulose in lithium chloride and N,N-dimethylacetamide.
McCormick CL, et al.
Macromolecules, 18(12), 2394-2401 (1985)
Hart WA, et al.
The Chemistry of Lithium, Sodium, Potassium, Rubidium, Cesium and Francium, 355-355 (1973)
Eagleson M.
Concise Encyclopedia Chemistry, 605-605 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service