Skip to Content
Merck
All Photos(1)

Key Documents

44213

Sigma-Aldrich

Thromboplastin from rabbit

Synonym(s):

Thromboplastin, Factor III, Thrombokinase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

biological source

rabbit

form

lyophilized solid

UniProt accession no.

storage temp.

2-8°C

Gene Information

rabbit ... F3(100009127)

Looking for similar products? Visit Product Comparison Guide

Application

Thromboplastin from rabbit has been used:
  • as a standard in tissue factor procoagulant activity assay (TF-PCA)
  • in pulmonary embolism assay
  • as a positive control in clotting assay using human plasma

Biochem/physiol Actions

Thromboplastin (Tissue factor) is a transmembrane protein, the chief in vivo activator of blood coagulation that leads to thrombin generation and fibrin deposition. It is also important in the activation of the coagulation protease cascade and is involved in the lethality of sepsis.,
Thromboplastin from rabbit might be used to produce disseminated intravascular coagulation (DIC) in animal models.

Packaging

One vial contains 3-4 mg thromboplastin.

Reconstitution

Reconstitute the content of 1 vial in 4 ml 10 mm CaCl2

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M E Bromberg et al.
Proceedings of the National Academy of Sciences of the United States of America, 92(18), 8205-8209 (1995-08-29)
Several studies have established a link between blood coagulation and cancer, and more specifically between tissue factor (TF), a transmembrane protein involved in initiating blood coagulation, and tumor metastasis. In the study reported here, a murine model of human melanoma
Line Olrik Berthelsen et al.
Thrombosis research, 126(4), 337-344 (2010-07-20)
Validation of animal models of disseminated intravascular coagulation (DIC) to human DIC is crucial in order to translate findings in research models to treatment modalities for DIC in humans. ISTH classifications of overt and non-overt human DIC have proven to
Esterina D'Asti et al.
Methods in molecular biology (Clifton, N.J.), 1458, 39-58 (2016-09-02)
Angiogenesis represents one aspect in the complex process that leads to the generation of the vascular tumor stroma. The related functional constituents include responses of endothelial, mural, bone marrow-derived, and resident inflammatory cells as well as activation of coagulation and
J Erlich et al.
Infection and immunity, 67(5), 2540-2546 (1999-05-04)
Tissue factor (TF) is the major activator of the coagulation protease cascade and contributes to lethality in sepsis. Despite several studies analyzing TF expression in animal models of endotoxemia, there remains debate about the cell types that are induced to
Nan Zhang et al.
The Journal of experimental medicine, 216(6), 1291-1300 (2019-05-03)
Macrophages resident in different organs express distinct genes, but understanding how this diversity fits into tissue-specific features is limited. Here, we show that selective expression of coagulation factor V (FV) by resident peritoneal macrophages in mice promotes bacterial clearance in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service