Skip to Content
Merck
All Photos(1)

Key Documents

32745

Sigma-Aldrich

Malachite Green oxalate salt

for microscopy, crystalline, S. No.: 754

Synonym(s):

N,N,N′,N′-Tetramethyl-4,4′-diaminotriphenylcarbenium oxalate, Basic Green 4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H25N2 · C2HO4 · 0.5C2H2O4
CAS Number:
Molecular Weight:
463.50
Colour Index Number:
42000
Beilstein:
3644550
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
MA.02
Pricing and availability is not currently available.

grade

for microscopy

Quality Level

form

crystalline

technique(s)

microbe id | staining: suitable

color

S. No.: 754

mp

144-150 °C

solubility

methanol: 0.01 g/10 mL, blue to very deep blue

εmax

≥1450 at 609-619 nm in water
≥300 at 420-430 nm in water

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

C[N+](C)=C1C=CC(C=C1)=C(C2=CC=C(N(C)C)C=C2)C3=CC=CC=C3.[O-]C(C(O)=O)=O

InChI

1S/C23H25N2.C2H2O4/c1-24(2)21-14-10-19(11-15-21)23(18-8-6-5-7-9-18)20-12-16-22(17-13-20)25(3)4;3-1(4)2(5)6/h5-17H,1-4H3;(H,3,4)(H,5,6)/q+1;/p-1

InChI key

REZFCYOVFCJTDG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Malachite green is manufactured as a zinc-free oxalate salt (green crystals with a metallic sheen) or as a zinc chloride salt (yellow crystals). Malachite green comes under the triphenyl methane class of dyes and is one of the most effective agents traditionally known for treating water mold infections of fish and eggs. It has also been found to be effective against ectoparasites, protozoan and some myxozoan parasites. Though it has several disadvantages like it acts as respiratory poison, teratogen and suspected carcinogen. It generated free radicals, thereby making it highly toxic to mammalian cells in culture; it also causes malignant transformation. It has a long half-life and can persist in tissues for long periods. In water, malachite green exists in equilibrium between the colored, ionic form and the colorless, nonionic pseudobase (carbinol form). Only the colored ionic form has antimicrobial activity, whereas the carbinol form is lipid-soluble, hence allowing it to pass across membranes. The toxicity of malachite green is dependent on temperature and it increase with increase in temperature, whereas it is more toxic at low pH and low hardness. It is phyotoxic in nature, thereby it is inactivated (oxidised) by light.[1]

Application

Malachite green oxalate salt was used in studying the photocatalytic degradation with TiO2/WO3.[2] It was also used in preparing selective medium for isolating Verticillium fungicola from mushroom.[3] It may be used for the determination of phosphate in serum.[4] It may also be used to monitor the presence of free COOH on the solid-phase.[5] It may be used as reference materials for in an analytical method to independently quantify malachite green oxalate and leucomalachite green in fish tissue.[6] It may also be used in Rappaport broth for incubating the membrane filters used in detecting Salmonella sp.[7]

Other Notes

The molecular mass of malachite green oxalate is expressed as either:
927.00 g/mol - the mass of 2 mol malachite green + 2 mol monohydrogen oxalate + 1 mol dihydrogen oxalate, or
463.50 g/mol - the mass of 1 mol malachite green + 1 mol monohydrogen oxalate + 0.5 mol dihydrogen oxalate

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Photocatalytic degradation of malachite green dyes with TiO2/WO3 composite.
Bojinova, Assya S., et al.
Eurasian Journal of Analytical Chemistry, 3, 34-43 (2008)
A Kallner
Clinica chimica acta; international journal of clinical chemistry, 59(1), 35-39 (1975-02-22)
The highly sensitive malachite-green method for the determination of phosphate requires a protective colloid in order to avoid precipitation of the formed dye salt. A polyvinylalcohol proved to be suitable and permit the determination of phosphate in urine and in
Malachite green, a valuable reagent to monitor the presence of free COOH on the solid-phase.
Attardi ME
Tetrahedron Letters, 41 (38), 7391-7394 (2000)
A selective medium for Verticillium fungicola.
Rinker, D. L., S. Bussmann, and G. Aim.
Canadian Journal of Plant Pathology, 15, 123-124 (1993)
M Gilgen et al.
Applied and environmental microbiology, 61(4), 1226-1231 (1995-04-01)
We have developed a simple, fast, and efficient procedure to detect enteroviruses in water samples. Aliquots of water are subjected to two-step filtration, with the second filter containing a positively charged nylon membrane that holds back virus particles. Viruses thus

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service