Skip to Content
Merck
All Photos(1)

Key Documents

90903

Sigma-Aldrich

Schaeffer and Fulton Spore Stain Solution A

for microscopy

Synonym(s):

Malachite Green Solution according to Schaeffer and Fulton

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12171500
NACRES:
MA.02

grade

for microscopy

product line

BioChemika

form

solution

shelf life

limited shelf life, expiry date on the label

technique(s)

microbe id | staining: suitable

suitability

spore forming microorganisms

SMILES string

[N+](=C4C=CC(=C(c6ccc(cc6)N(C)C)c5ccccc5)C=C4)(C)C.[N+](=C1C=CC(=C(c3ccc(cc3)N(C)C)c2ccccc2)C=C1)(C)C.[O-]C(=O)C(=O)[O-].OC(=O)C(=O)O.OC(=O)C(=O)O

InChI

1S/2C23H25N2.3C2H2O4/c2*1-24(2)21-14-10-19(11-15-21)23(18-8-6-5-7-9-18)20-12-16-22(17-13-20)25(3)4;3*3-1(4)2(5)6/h2*5-17H,1-4H3;3*(H,3,4)(H,5,6)/q2*+1;;;/p-2

InChI key

CNYGFPPAGUCRIC-UHFFFAOYSA-L

General description

In Schaeffer-Fulton`s method, a primary stain-malachite green is forced into the spore by steaming the bacterial emulsion. Malachite green is water soluble and has a low affinity for cellular material, so vegetative cells may be decolorized with water. Vegetative cells are then counterstained with safarin (Schaeffer & Fulton`s Spore Stain B, Cat. No. 39955). The components and the method are also available as kit (Cat. No. 04551).

Components

Composition:
Schaeffer & Fulton`s Spore Stain A: Malachite green 50 g/l in water

replaced by

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Repr. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia Zeng et al.
Biophysical journal, 104(1), 139-145 (2013-01-22)
It is shown that the nonlinear optical phenomenon known as second-harmonic generation can be used for label-free, time-resolved study of the transport of molecules through living cell membranes. The adsorption and transport of a 300-Da molecular-mass hydrophobic ion at the
Xiaofang Jia et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(42), 13494-13500 (2012-09-05)
This work describes a novel strategy for the highly sensitive and selective detection of cysteine (Cys) and glutathione (GSH) based on the Hg(2+)-AGRO100-malachite green (MG) complex system. The dye MG, which has a very low quantum yield in aqueous solution
Ola M Gomaa et al.
Cell biochemistry and biophysics, 65(3), 413-423 (2012-10-19)
Filamentous fungi show great promise in remediation of environmental contaminants such as industrial dyes. In the current study, Aspergillus niger (Genbank ID: JF437542) decolorized 82 % of the test dye malachite green (MG; 50 mg/l) during cultivation for 24 h.
P Manoj Kumar Reddy et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 67(5), 1097-1104 (2013-02-19)
Oxidative decomposition of aqueous organic pollutant malachite green (MG) was studied in a dielectric barrier discharge reactor operated under ambient conditions. Total organic carbon content analysis confirmed the mineralization of the pollutant leading to the formation of carbon dioxide, which
R Sahraei et al.
Environmental monitoring and assessment, 185(7), 5817-5822 (2012-12-04)
A new spectrophotometric method is reported for the determination of nanomolar level of malachite green in surface water samples. The method is based on the catalytic effect of silver nanoparticles on the oxidation of malachite green by hexacyanoferrate (III) in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service