Skip to Content
Merck
All Photos(1)

Key Documents

310M-2

Sigma-Aldrich

Parathyroid Hormone (PTH) (MRQ-31) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

MRQ-31, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (310M-24)
vial of 0.5 mL concentrate (310M-25)
bottle of 1.0 mL predilute (310M-27)
vial of 1.0 mL concentrate (310M-26)
bottle of 7.0 mL predilute (310M-28)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG2a

control

parathyroid tissue

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... PTH(5741)

General description

The rate of parathormone secretion is directly responsive to the level of calcium in the serum, and indeed the cytoplasm, of parathyroid cells, as has been shown by studies both in vivo and in vitro (Brown et al, 1982). Recent in vitro studies of osteoclast turnover suggest that both PTH and PTH-related protein exert both pro- and anti-apoptotic effects in mesenchymal cells (Chen et al, 2002)

Surgical pathologists are familiar with the ability of parathyroid proliferations to assume a variety of histological guises, posing difficulty to categorize any given lesion as hyperplastic, adenomatous or carcinomatous in nature (Wick et al, 1997). This is usually resolved with macroscopic appearance of the remaining parathyroid glands as assessed by the surgeon. The role of the surgical pathologist is to identify the lesion as parathyroid in nature and to assess whether it is normocellular or hypercellular. Although easily accomplished in the majority of instances, rare examples of parathyroid hyperplasia/adenoma showing a follicular/trabecular arrangement may cause concern over the alternative diagnosis of a thyroid adenoma. This becomes more pertinent when the parathyroid lesion abuts into the thyroid gland or lies within the thyroid capsule. Immunostaining for thyroglobulin and parathyroid hormone (PTH) is especially useful to resolve the problem (Permanetter et al, 1983). Nevertheless, caution should be exercised since parathyroid cells often discharge their hormonal product almost as soon as it is packaged in the cytoplasm, resulting in false-negative anti-PTH immunostaining, although the cells are biologically synthetic (Wick et al, 1997)

Anti-PTH antibody is also useful to distinguish parathyroid hyperplasia/neoplasms from thyroid and metastatic neoplasms (Wick et al, 1997); although the pathologist is typically aware of the preoperative hypercalcemic status. Occasionally when the surgeon does not supply this information PTH immunohistochemistry is essential. Even more problematic, are situations in which clear cell parathyroid carcinomas are nonsecretory without an abnormality in mineral metabolism (Aldinger et al, 1982). In such situations, metastatic renal cell carcinoma or metastatic clear cell carcinoma of the lung is evident warranting PTH immunohistochemistry to arrive at the correct diagnosis (Wick et al, 1997). The other instance in which anti-PTH antibodies are useful is in the consideration of parathyroid carcinomas located primarily in the anterior mediastinum (intrathymically). In this situation distinction from primary thymic metastatic carcinomas, non-Hodgkin′s lymphoma and germ cell tumors is necessary (Murphy et al, 1986).

The diagnosis of the majority of parathyroid proliferation may be accomplished with an adequate history, biochemistry profile, and histomorphological assessment; however, rare instances in which the tumors have an abnormal location, clear cell morphology, or a non-secretory may result in erroneous diagnoses, warranting anti-PTH immunohistochemistry.

Quality


IVD

IVD

IVD

RUO

Linkage

Parathyroid Hormone (PTH) Positive Control Slides, Product No. 310S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Parathyroid hormone: biochemical aspects of biosynthesis, secretion, action, and metabolism.
J F Habener et al.
Physiological reviews, 64(3), 985-1053 (1984-07-01)
M N Murphy et al.
Cancer, 58(11), 2468-2476 (1986-12-01)
Nonsecretory parathyroid carcinoma is rare, particularly in extracervical sites. The authors present a case of a 51-year-old man with a large mediastinal mass that was found on a chest x-ray. Light and electron microscopy and immunohistochemical analysis of the resected
N G Ordoñez et al.
The American journal of surgical pathology, 7(6), 535-542 (1983-09-01)
Nonfunctioning carcinomas of the parathyroid gland are rare and difficult to diagnose. They are often confused with thyroid tumors or with metastasis from other sites. We report two cases of nonfunctioning parathyroid carcinomas; one was originally diagnosed as follicular carcinoma
K A Aldinger et al.
Cancer, 49(2), 388-397 (1982-01-15)
Seven cases of functioning and two cases of nonfunctioning parathyroid carcinoma are presented together with a general discussion of this disease. Functioning parathyroid carcinoma may be associated with prolonged survival. The morbidity and mortality from this disease are generally related
M R Wick et al.
Seminars in diagnostic pathology, 14(3), 183-202 (1997-08-01)
Clear cell tumors of the endocrine system and thymus are rare lesions with which surgical pathologists have only infrequent contact. As a result, these neoplasms may be misinterpreted and incorrectly classified. This review considers the histological, immunohistochemical, and ultrastructural features

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service