Skip to Content
Merck
All Photos(1)

Documents

226R-1

Sigma-Aldrich

BCL2 (E17) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

E17, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (226R-14)
vial of 0.5 mL concentrate (226R-15)
bottle of 1.0 mL predilute (226R-17)
vial of 1.0 mL concentrate (226R-16)
bottle of 7.0 mL predilute (226R-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG

control

tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... BCL2(596)

General description

Anti-BCL2 has shown consistent negative reaction on reactive germinal centers and positive staining of neoplastic follicles in follicular lymphoma. Consequently, this antibody is valuable when distinguishing between reactive and neoplastic follicular proliferation in lymph node biopsies. This antibody may also be used in distinguishing between those follicular lymphomas that express BCL2 protein and the small number in which the neoplastic cells are BCL2 negative.

Linkage

BCL2 Positive Control Slides, Product No. 226S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Tsujimoto et al.
Proceedings of the National Academy of Sciences of the United States of America, 83(14), 5214-5218 (1986-07-01)
We have determined that the bcl-2 (B-cell leukemia/lymphoma 2) gene is transcribed into three overlapping mRNAs, and we have cloned bcl-2 cDNA sequences. Sequence analysis of the bcl-2 cDNA clones and comparison of their sequences to their genomic counterparts indicate
D Hockenbery et al.
Nature, 348(6299), 334-336 (1990-11-22)
The t(14; 18) chromosomal translocation of human follicular B-cell lymphoma juxtaposes the bcl-2 gene with the immunoglobulin heavy chain locus. The bcl-2 immunoglobulin fusion gene is markedly deregulated resulting in inappropriately elevated levels of bcl-2 RNA and protein. Transgenic mice
F Pezzella et al.
The American journal of pathology, 137(2), 225-232 (1990-08-01)
It has been reported previously that the bcl-2 protooncogene protein is detectable in neoplastic cells from cases of human lymphoma in which the 14;18 chromosomal translocation is present, but not in lymphomas that lack this chromosomal rearrangement or in normal
D R Ciocca et al.
Endocrine, 13(1), 1-10 (2000-10-29)
Tamoxifen is one of the most effective treatments for breast cancer. Standard practice is to select patients who are likely to respond to this therapy through the evaluation of estrogen receptor (ER) and progesterone receptor (PR) in the primary tumor
B Martin et al.
British journal of cancer, 89(1), 55-64 (2003-07-03)
The role of the anti-apoptotic protein Bcl-2 in lung cancer remains controversial. In order to clarify its impact on survival in small and non-small cell lung cancer (NSCLC), we performed a systematic review of the literature. Trials were selected for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service