All Photos(1)
About This Item
Linear Formula:
K14CN
CAS Number:
Molecular Weight:
65.12
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.25
grade:
for analytical purposes
Recommended Products
grade
for analytical purposes
Quality Level
extent of labeling
10-15 mCi per mmol
mass shift
M+2
storage temp.
2-8°C
SMILES string
[K][14CH2]C#N
InChI
1S/C2H2N.K/c1-2-3;/h1H2;/i1+2;
InChI key
IJAJWWZKTPVZIH-DEQYMQKBSA-N
Looking for similar products? Visit Product Comparison Guide
Packaging
Packaged in screw-cap bottle
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Xiao-Zhang Yu et al.
Journal of hazardous materials, 225-226, 190-194 (2012-05-29)
A study was conducted to investigate activities of nitrate reductase (NR) and glutamine synthetase (GS) in plants during cyanide metabolism. Young rice seedlings (Oryza sativa L. cv. XZX 45) were grown in the nutrient solutions containing KNO(3) or NH(4)Cl and
Ruth Chang et al.
Plant molecular biology, 78(1-2), 109-122 (2011-11-17)
Mitogen-activated protein kinases (MPKs) are regulated by diverse stresses with a reactive oxygen species (ROS) component. Here, we report the rapid and transient activation of MPK3, MPK4 and MPK6 upon oxygen deprivation as well as reoxygenation in seedlings of Arabidopsis
Erica M Granjeiro et al.
Autonomic neuroscience : basic & clinical, 167(1-2), 21-26 (2011-12-27)
Several studies from our group have indicated that the BNST play an important role in baroreflex modulation. However, the involvement of the BNST in the chemoreflex activity is unknown. Thus, in the present study, we investigated the effect of the
Anand Kumar et al.
Amino acids, 43(6), 2417-2429 (2012-05-23)
Condensation reactions of amino acid (glycine and alanine) on the surface of metal(II) octacyanomolybdate(IV) (MOCMo) complexes are investigated using high-performance liquid chromatography (HPLC) and electron spray ionizations-mass spectroscopy (ESI-MS). The series of MOCMo have been synthesized and the effect of
Wenying Jian et al.
Journal of the American Society for Mass Spectrometry, 23(5), 964-976 (2012-03-07)
Drugs can be metabolically activated to soft and hard electrophiles, which are readily trapped by glutathione (GSH) and cyanide (CN), respectively. These adducts are often detected and structurally characterized using separate tandem mass spectrometry methods. We describe a new method
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service