Skip to Content
Merck
All Photos(1)

Key Documents

60178

Sigma-Aldrich

Potassium cyanide

BioUltra, ≥98.0% (AT)

Synonym(s):

KCN, Potassium salt of hydrocyanic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KCN
CAS Number:
Molecular Weight:
65.12
Beilstein:
3593645
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

product line

BioUltra

Assay

≥98.0% (AT)

form

crystalline

impurities

insoluble matter, passes filter test

pH

11.0-13.0 (25 °C, 1 M in H2O)

mp

634 °C (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤200 mg/kg
sulfate (SO42-): ≤400 mg/kg
sulfide (S2-): ≤5 mg/kg
thiocyanate (SCN-): ≤100 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤100 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤5000 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.06
λ: 280 nm Amax: 0.03

SMILES string

[K]C#N

InChI

1S/CN.K/c1-2;

InChI key

YUZRZFQHUCKACF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Potassium cyanide is use to inhibit oxidative phosphorylation reactions in a wide variety of biological systems and applications.

Other Notes

Blocks electron transport to oxygen via cytochrome oxidase, hence employed in the determination of succinate dehydrogenase in submitochondrial particles by preventing substantial loss of succinate by electron flux to O2[1][2]

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Met. Corr. 1 - STOT RE 1

Target Organs

Thyroid

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pascaline Clerc et al.
Journal of bioenergetics and biomembranes, 39(2), 158-166 (2007-04-21)
We have studied the effect of nitric oxide (NO) and potassium cyanide (KCN) on oxidative phosphorylation efficiency. Concentrations of NO or KCN that decrease resting oxygen consumption by 10-20% increased oxidative phosphorylation efficiency in mitochondria oxidizing succinate or palmitoyl-L-carnitine, but
EPR and other properties of succinate dehydrogenase.
T Ohnishi et al.
Methods in enzymology, 53, 483-495 (1978-01-01)
Yudong Wang et al.
The Journal of biological chemistry, 285(39), 29834-29841 (2010-07-29)
Fatty acid β-oxidation (FAO) and oxidative phosphorylation (OXPHOS) are key pathways involved in cellular energetics. Reducing equivalents from FAO enter OXPHOS at the level of complexes I and III. Genetic disorders of FAO and OXPHOS are among the most frequent
Mammalian succinate dehydrogenase.
B A Ackrell et al.
Methods in enzymology, 53, 466-483 (1978-01-01)
Takeshi Hatani et al.
Biochemical and biophysical research communications, 505(4), 1251-1256 (2018-10-20)
Many studies have shown the feasibility of in vivo cardiac transplantation of human induced pluripotent stem cell-derived cardiomyocytes (iPSC-CMs) in animal experiments. However, nano-structural confirmation of the successful incorporation of the engrafted iPSC-CMs including electron microscopy (EM) has not been accomplished

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Hello, please can this type be used for laboratory experiments on experimental rodents 

    1 answer
    1. Product No. 60178 is not intended for live animal testing.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service