Skip to Content
Merck
All Photos(1)

Key Documents

143M-1

Sigma-Aldrich

CD43 (MT1) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

MT1, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (143M-14)
vial of 0.5 mL concentrate (143M-15)
bottle of 1.0 mL predilute (143M-17)
vial of 1.0 mL concentrate (143M-16)
bottle of 7.0 mL predilute (143M-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:200

isotype

IgG1

control

tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

membranous

Gene Information

human ... SPN(6693)

General description

From 70 to 90% of T-cell Lymphomas and from 22 to 37% of B cell Lymphomas are labeled with this antibody. No reactivity has been observed with reactive B cells. So, a B lineage population that co-expresses a T-cell marker is highly likely to be a malignant lymphoma rather than a reactive B cell population. When used in combination with CD45 and L26, effective immunophenotyping of the majority of paraffin-embedded lymphomas can be obtained. Co-staining of a lymphoid infiltrate with CD20 and CD3 argues against a reactive process and favors lymphoma.

Quality


IVD

IVD

IVD

RUO

Linkage

CD43 Positive Control Slides, Product No. 143S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arber, DA, et al.
Appl. Immunohistochem., 1, 88-96 (1993)
J G Strickler et al.
Human pathology, 18(8), 808-814 (1987-08-01)
The immunoreactivity of eight monoclonal antibodies was evaluated on 45 routinely processed lymphomas (22 T-cell lymphomas, 11 B-cell lymphomas, and 12 cases of Hodgkin's disease). Two antibodies reactive with leukocyte common (T200) antigens (PD7/26 and 2B11) stained most of the
K Sheibani et al.
Human pathology, 18(10), 1051-1062 (1987-10-01)
From the study of over 3,000 immunologically well characterized cases, we have developed a systematic approach for the immunohistologic evaluation of lymphoproliferative disorders (LPD). By using a basic screening panel of antibodies on fresh-frozen sections, LPD can be reliably immunophenotyped.
J K Chan et al.
Histopathology, 12(5), 461-480 (1988-05-01)
This review aims to provide a simple guide and quick reference to the terminology and diagnostic applications of leucocyte monoclonal antibodies. The differentiation cluster terminology, where applicable, is used throughout.
J M Cabeçadas et al.
Histopathology, 19(5), 419-424 (1991-11-01)
Five antibodies, MT1 (CD43), UCHL1 (CD45RO), OPD4, poly-CD3 and beta F1, were assessed for their reactivity with 50 archival cases of T-cell lymphoma in formalin-fixed paraffin-embedded tissue. All cases had been previously characterized as T-cell lymphomas, and the histological types

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service