Skip to Content
Merck
All Photos(1)

Key Documents

T7209

Sigma-Aldrich

1,1,1,2-Tetrachloroethane

ReagentPlus®, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CCl3
CAS Number:
Molecular Weight:
167.85
Beilstein:
1733216
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.04
grade:
reagent
bp:
138 °C (lit.)
vapor pressure:
12.0 mmHg

grade

reagent

vapor pressure

12.0 mmHg

product line

ReagentPlus®

Assay

99%

form

liquid

dilution

(for general lab use)

refractive index

n20/D 1.481 (lit.)

bp

138 °C (lit.)

mp

-70.2 °C

density

1.598 g/mL at 25 °C (lit.)

SMILES string

ClCC(Cl)(Cl)Cl

InChI

1S/C2H2Cl4/c3-1-2(4,5)6/h1H2

InChI key

QVLAWKAXOMEXPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A Thompson et al.
Chemico-biological interactions, 51(3), 321-333 (1984-10-01)
The susceptibility of polychlorinated ethanes to reductive metabolism was evaluated by measuring the amount of each compound consumed during anaerobic incubations with rat live microsomes; 1,1,1,2-tetrachloroethane, pentachloroethane and hexachloroethane were metabolized extensively, 1,1,1,2-tetrachloroethane and the trichloroethanes were metabolized very slowly
L O Svaasand et al.
Lasers in surgery and medicine, 32(2), 137-142 (2003-02-01)
Cryogen spray cooling (CSC) is used extensively for epidermal protection during laser-induced photothermolysis of port wine stains and other vascular skin lesions. The efficacy of CSC depends critically on the heat transfer coefficient (H) at the skin surface for which
D L Story et al.
Toxicology and industrial health, 2(4), 351-362 (1986-12-01)
Nine chlorinated aliphatics (CAs)--1,1-dichloroethane, 1,2-dichloroethane, 1,1,1-trichloroethane, 1,1,2-trichloroethane, trichloroethylene, tetrachloroethylene, 1,1,1,2-tetrachloroethane, 1,1,2,2-tetrachloroethane, and hexachloroethane--were examined in a rat liver foci assay for evidence of initiating and promoting potential. Young adult male Osborne-Mendel rats (ten/group) were given partial hepatectomies, followed 24 hr
M Paolini et al.
Mutagenesis, 5(1), 51-54 (1990-01-01)
The synergistic effect of NADPH and NADH on P450 functions upon pre-mutagens requiring metabolism during the incubation conditions used in the liver microsomal assay (LMA) was studied. The mean specific activity (Asp) during 1 h of pre-incubation (LMA) of some
1,1,1,2-Tetrachloroethane.
IARC monographs on the evaluation of the carcinogenic risk of chemicals to humans, 41, 87-97 (1986-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service