Skip to Content
Merck
All Photos(2)

Key Documents

C3654

Sigma-Aldrich

Ammonium cerium(IV) nitrate

≥98% (titration)

Synonym(s):

Ceric ammonium nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ce(NH4)2(NO3)6
CAS Number:
Molecular Weight:
548.22
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55

Quality Level

Assay

≥98% (titration)

form

powder or crystals

reaction suitability

reagent type: oxidant

pH

0.61 (20 °C, 111.11 g/L)

SMILES string

N.N.[Ce+4].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Ce.2HNO3.4NO3.2H3N/c;6*2-1(3)4;;/h;2*(H,2,3,4);;;;;2*1H3/q+4;;;4*-1;;

InChI key

WIBGOERAEYJBOT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 2 - Skin Corr. 1B - Skin Sens. 1A

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Sudha et al.
Ultrasonics sonochemistry, 19(5), 994-998 (2012-03-09)
Tetrahydrobenzo[c]xanthenes-11-ones was synthesized by a three component reaction of α-naphthol, aromatic aldehyde and dimedone. Ceric ammonium nitrate acts as a suitable eco-friendly catalyst for this method. Shorter reaction duration, mild reaction condition and low cost make this protocol more effective.
Weibin Song et al.
Bioorganic & medicinal chemistry letters, 22(1), 387-390 (2011-11-26)
A new, practical and efficient method for the synthesis of anhydrovinblastine AVBL (1f) by oxidative coupling of vindoline and catharanthine in the presence of ceric ammonium nitrate (CAN) was developed. Under the optimized reaction conditions, we synthesized a new series
D Subhas Bose et al.
Journal of combinatorial chemistry, 12(1), 100-110 (2009-12-17)
An efficient and practical method has been manifested for the diversity-oriented synthesis of quinolines via Friedländer annulation reaction for the generation of a wide range of structurally interesting and pharmacologically significant compounds by using ceric ammonium nitrate as a catalyst
Rasika Tankhiwale et al.
Colloids and surfaces. B, Biointerfaces, 69(2), 164-168 (2009-01-10)
The present work describes ceric ammonium nitrate (CAN) initiated graft copolymerization of acrylamide onto cellulose-based filter paper followed by entrapment of silver nanoparticles. The copolymerization was carried out in aqueous solution, containing 2M acrylamide monomer and 16mM N,N'-methylene bisacrylamide (MB)
David Kesselring et al.
Journal of the American Chemical Society, 130(34), 11290-11291 (2008-08-06)
Conditions for a site-selective ceric ammonium nitrate oxidation have been developed. The reactions proceed nicely on both 1K- and 12K-microelectrode arrays. The procedure for developing the reactions was very simple and demonstrated that the same reagents used for a solution-phase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service