Skip to Content
Merck
All Photos(1)

Key Documents

83379

Sigma-Aldrich

Mercury(II) iodide red

puriss. p.a., ACS reagent, red, ≥99.0% (RT)

Synonym(s):

Mercuric iodide red

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HgI2
CAS Number:
Molecular Weight:
454.40
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24
form:
powder

grade

ACS reagent
puriss. p.a.

Quality Level

Assay

≥99.0% (RT)

form

powder

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤20 mg/kg
Hg+: ≤1000 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

I[Hg]I

InChI

1S/Hg.2HI/h;2*1H/q+2;;/p-2

InChI key

YFDLHELOZYVNJE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Mercury(II) iodide red (HgI2) is formed during iodometric analysis.

Application

Mercury(II) iodide may be used in the preparation of mercury(II) acetate and di­iodo­bis(1-methyl-1,3-imidazolium-2-thiol­ato-S)­mercury(II).

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diiodobis (1-methyl-1, 3-imidazolium-2-thiolato-S) mercury (II).
Pavlovic G, et al.
Acta Crystallographica Section C, Crystal Structure Communications, 56(7), 801-803 (2000)
Eagleson M.
Concise Encyclopedia Chemistry, 637-637 (1994)
Health hazard from mercury soap.
E S Mucklow
Lancet (London, England), 1(8635), 448-448 (1989-02-25)
Zhong Su et al.
Physics in medicine and biology, 50(12), 2907-2928 (2005-06-03)
The signal properties of polycrystalline mercuric iodide (HgI2) film detectors, under irradiation conditions relevant to mammographic, radiographic, fluoroscopic and radiotherapy x-ray imaging, are reported. Each film detector consists of an approximately 230 to approximately 460 microm thick layer of HgI2
[Experimental study of the allergenic activity of red mercuric iodide as an atmospheric pollutant].
A B Ermachenko
Gigiena i sanitariia, (11)(11), 82-83 (1983-11-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service