Skip to Content
Merck
All Photos(1)

Key Documents

221228

Sigma-Aldrich

Ammonium hydroxide solution

ACS reagent, 28.0-30.0% NH3 basis

Synonym(s):

Ammonia aqueous, Ammonia water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4OH
CAS Number:
Molecular Weight:
35.05
Beilstein:
3587154
MDL number:
UNSPSC Code:
12352106
eCl@ss:
38060103
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

form

liquid

autoignition temp.

1204 °F

expl. lim.

27 %

concentration

28.0-30.0% NH3

impurities

MnO4- reducers, passes test
≤0.002% CO2

ign. residue

≤0.002%

pH

11.7 (20 °C)

density

0.9 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.5 ppm
nitrate (NO3-): ≤2 ppm
phosphate (PO43-): ≤2 ppm
sulfate (SO42-): ≤2 ppm

cation traces

Fe: ≤0.2 ppm
heavy metals: ≤0.5 ppm (by ICP)

storage temp.

2-8°C

SMILES string

[NH4+].[OH-]

InChI

1S/H3N.H2O/h1H3;1H2

InChI key

VHUUQVKOLVNVRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium hydroxide is a weak base mainly used as a source to prepare other ammonium compounds like urea, ammonium carbonate and ammonium sulfate etc.

Application

Ammonium hydroxide solution may be used:
  • As a medium to prepare magnetite (Fe3O4) nanoparticles via a modified controlled chemical coprecipitation method.
  • As a solvent and reagent to prepare 4-aryl and 4-alkyl-2,6-dimethyl-1,4-dihydro-3,5-pyridinedicarboxylates via Hantzsch synthesis in a microwave cavity synthesizer.

Other Notes

ACS specifications same for product 320145

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christie Ying Kei Lung et al.
Dental materials journal, 32(1), 165-172 (2013-02-02)
The effect of silica-coating by sol-gel process on the bond strength of resin composite to zirconia was evaluated and compared against the sandblasting method. Four groups of zirconia samples were silica-coated by sol-gel process under varied reagent ratios of ethanol
An efficient and fast procedure for the Hantzsch dihydropyridine synthesis under microwave conditions.
Oehberg L and Westman J
Synlett, 2001(08), 1296-1298 (2001)
Synthesis and characterization of biocompatible Fe3O4 nanoparticles.
Sun J, et al.
Journal of Biomedical Materials Research Part A, 80(2), 333-341 (2007)
Ramakrishna A
Goyal?s IIT foundation course chemistry: For Class-9, 160-160 (2014)
Isolation and characterization of mesenchymal stem cells.
Odabas SA, et al.
Bone Marrow and Stem Cell Transplantation., 47-63 (2014)

Articles

Acid and base chart lists the strength of acids and bases (strongest to weakest) in order. Simple to use laboratory reference chart for scientists, researchers and lab technicians.

Acid and base chart lists the strength of acids and bases (strongest to weakest) in order. Simple to use laboratory reference chart for scientists, researchers and lab technicians.

Acid and base chart lists the strength of acids and bases (strongest to weakest) in order. Simple to use laboratory reference chart for scientists, researchers and lab technicians.

Acid and base chart lists the strength of acids and bases (strongest to weakest) in order. Simple to use laboratory reference chart for scientists, researchers and lab technicians.

See All

Protocols

In the present study we demonstrate compliance of silica gel 60 F254 HPTLC plates with USP requirements, and how HPTLC plates are suitable for the Sildenafil citrate-Limit of Imidazole test.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service