Skip to Content
Merck
All Photos(1)

Key Documents

71360

Sigma-Aldrich

Sodium carbonate decahydrate

puriss. p.a., ≥99.0% (T)

Synonym(s):

Sal soda, Soda

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2CO3 · 10H2O
CAS Number:
Molecular Weight:
286.14
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.0% (T)
form:
crystals
Pricing and availability is not currently available.

grade

puriss. p.a.

Quality Level

Assay

≥99.0% (T)

form

crystals

impurities

≤0.0005% total nitrogen (N)
≤0.003% total sulfur (as SO4)

pH

11-12 (25 °C, 50 g/L)

anion traces

chloride (Cl-): ≤5 mg/kg
phosphate, silicate (as SiO2): ≤20 mg/kg

cation traces

Al: ≤5 mg/kg
Ca: ≤20 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤2 mg/kg
K: ≤50 mg/kg
Mg: ≤2 mg/kg
Mn: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]C([O-])=O

InChI

1S/CH2O3.2Na.10H2O/c2-1(3)4;;;;;;;;;;;;/h(H2,2,3,4);;;10*1H2/q;2*+1;;;;;;;;;;/p-2

InChI key

XYQRXRFVKUPBQN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Sodium carbonate decahydrate is an organooxygen compound. It is used as a strong base or a powerful electrolyte.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dehydration of sodium carbonate decahydrate to monohydrate in a fluidized bed.
Hartman M, et al.
AIChE Journal, 47(10), 2333-2333 (2001)
Chemical structure of poly (β-cyclodextrin-co-citric acid).
Bednarz S, et al.
Journal of Applied Polymer Science, 119(6), 3511-3520 (2011)
An infra-red study of the hydrates of sodium carbonate.
Buijs K and Schutte CJH.
Spectrochimica Acta Part A: Molecular Spectroscopy, 17(9), 917-920 (1961)
Azadeh Nilghaz et al.
Lab on a chip, 12(1), 209-218 (2011-11-18)
This paper describes the fabrication of microfluidic cloth-based analytical devices (μCADs) using a simple wax patterning method on cotton cloth for performing colorimetric bioassays. Commercial cotton cloth fabric is proposed as a new inexpensive, lightweight, and flexible platform for fabricating
Feng Gu et al.
Bioresource technology, 124, 299-305 (2012-09-20)
Green liquor consists of sodium carbonate and sodium sulfide and is readily available in any kraft mills. The green liquor pretreatment process for bioethanol production was developed for wood chips. This process uses only proven technology and equipment currently used

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service