Skip to Content
Merck
All Photos(1)

Key Documents

234591

Sigma-Aldrich

Methyl laurate

99.5%

Synonym(s):

Lauric acid methyl ester, Methyl dodecanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10CO2CH3
CAS Number:
Molecular Weight:
214.34
Beilstein:
1767780
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99.5%

form

liquid

refractive index

n20/D 1.431 (lit.)

bp

262 °C/766 mmHg (lit.)

mp

4-5 °C (lit.)

solubility

water: insoluble 0.00759 g/L at 25 °C (practically)

density

0.87 g/mL at 25 °C (lit.)

food allergen

no known allergens

functional group

ester

SMILES string

CCCCCCCCCCCC(=O)OC

InChI

1S/C13H26O2/c1-3-4-5-6-7-8-9-10-11-12-13(14)15-2/h3-12H2,1-2H3

InChI key

UQDUPQYQJKYHQI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydrogenation of methyl laurate catalyzed by diatomite supported Pd-M (M=Cu, Co, Ni) bimetal nanocatalysts has been reported. IR absorption spectrum of methyl laurate in carbon tetrachloride and carbon disulphide solutions has been compared with the spectra of derivatives deuterated in the ω-methyl group and α-methylene groups.

Application

Methyl laurate has been used in selective synthesis of the secondary amide surfactant, N-methyl lauroylethanolamide.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

282.2 °F - closed cup

Flash Point(C)

139 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Lipok et al.
Molecules (Basel, Switzerland), 25(14) (2020-07-28)
A closed atmospheric pressure chemical ionization (APCI) ion source as interface between a gas chromatograph (GC) and a triple quadrupole mass spectrometer (QqQ-MS) was developed. The influence of different ion source conditions, such as humidity, make-up gas flow, and the
The infrared absorption spectra of deuterated esters: III. Methyl laurate.
Jones RN.
Canadian Journal of Chemistry, 40(2), 301-320 (1962)
Y Komata et al.
Biological & pharmaceutical bulletin, 17(5), 705-708 (1994-05-01)
The effect of long chain fatty acid (FA) and its analogs on the accumulation of thiamine disulfide (TDS) in rat skin using propylene glycol as a vehicle was studied in vitro. Lauric acid (12:0) increased the accumulation of TDS in
E Barboza-Silva et al.
Oral microbiology and immunology, 24(4), 265-271 (2009-07-04)
The arginine deiminase system (ADS) of oral bacteria is a major generator of alkali (ammonia) in dental plaque and is considered to have anticaries effects. However, many of the antimicrobial agents used in oral care products may reduce alkali production
D A Dimas et al.
AAPS PharmSciTech, 1(2), E16-E16 (2004-01-20)
The effects of coating thickness, type of adhesive, and type and concentration of enhancer on the mechanical properties of two acrylic pressure-sensitive adhesives (PSAs) were investigated using a 2(4) factorial design and an optimization technique. Sixteen formulations containing 0% or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service