Skip to Content
Merck
All Photos(2)

Key Documents

61148

Sigma-Aldrich

Copper(II) acetate monohydrate

greener alternative

puriss. p.a., ≥99.0% (RT)

Synonym(s):

Cupric acetate monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cu(CO2CH3)2 · H2O
CAS Number:
Molecular Weight:
199.65
Beilstein:
3730548
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.21
form:
powder or crystals

vapor density

6.8 (vs air)

grade

puriss. p.a.

Assay

≥99.0% (RT)

form

powder or crystals

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤0.01% total nitrogen (N)

pH

5.2-5.5 (20 °C, 20 g/L)

anion traces

chloride (Cl-): ≤10 mg/kg
sulfate (SO42-): ≤500 mg/kg

cation traces

Ca: ≤20 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤20 mg/kg
Zn: ≤50 mg/kg

greener alternative category

SMILES string

O.CC(=O)O[Cu]OC(C)=O

InChI

1S/2C2H4O2.Cu.H2O/c2*1-2(3)4;;/h2*1H3,(H,3,4);;1H2/q;;+2;/p-2

InChI key

NWFNSTOSIVLCJA-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Copper(II) acetate monohydrate is a bivalent complex of copper. Its magnetic moment has been reported to be 1.4Bohr magnetons at room temperature. Its thermal degradation has been investigated by TG-DTG/DTA (Thermogravimetry-Derivative Thermogravimetry/Differential Thermal Analysis), DSC (Differential Scanning Calorimetry) and XRD (X-ray Diffraction) techniques.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalysis. Click here for more information.

Application


  • Impact of temperature on the physicochemical, structural and biological features of copper-silica nanocomposites.: This study investigates how temperature variations affect the properties of copper-silica nanocomposites. The research focuses on the physicochemical and structural changes that occur and their subsequent impact on biological applications. The findings provide valuable insights for the development of advanced materials with tailored properties for specific applications (Dulski et al., 2020).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The kinetics of thermal dehydration of copper (II) acetate monohydrate in air.
Zhang K, et al.
Thermochimica Acta, 437(1), 145-149 (2005)
Dinuclear copper (II) acetate monohydrate: a redetermination of the structure by neutron-diffraction analysis.
Brown GM and Chidambaram R.
Acta Crystallographica Section B, Structural Science, 29(11), 2393-2403 (1973)
F Heidenau et al.
Journal of materials science. Materials in medicine, 16(10), 883-888 (2005-09-17)
Postoperative implant-associated infection is still an unresolved and serious complication in modern surgery. Antibacterial and biocompatible surfaces could both reduce infection rates and promote tissue integration. In this respect, a comparative study of the antibacterial as well as the biocompatible
Alexander Navarrete et al.
Faraday discussions, 183, 249-259 (2015-09-24)
A novel plasmonic reactor concept is proposed and tested to work as a visible energy harvesting device while allowing reactions to transform CO2 to be carried out. Particularly the reverse water gas shift (RWGS) reaction has been tested as a
Łukasz Orzeł et al.
Dalton transactions (Cambridge, England : 2003), 44(13), 6012-6022 (2015-02-28)
The nature of chlorophyll interactions with copper(II) ions varies considerably in organic solvents, depending on the dominant coordinative form. Besides formation of the metallo tetrapyrrolic complex, Cu(II) ions can cause oxidation of the pigment, reversible or irreversible, which can lead

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service