Skip to Content
Merck
All Photos(4)

Documents

217557

Sigma-Aldrich

Copper(II) acetate monohydrate

greener alternative

ACS reagent, ≥98%

Synonym(s):

Cupric acetate monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cu(CO2CH3)2 · H2O
CAS Number:
Molecular Weight:
199.65
Beilstein:
3730548
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent

Quality Level

vapor density

6.8 (vs air)

Assay

≥98%
98.0-102.0% (ACS specification)

form

crystalline powder
lumps and powder

reaction suitability

reagent type: catalyst
core: copper

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤0.01% insolubles

pH

5.2-5.5 (20 °C, 20 g/L)

anion traces

chloride (Cl-): ≤0.003%
sulfate (SO42-): ≤0.01%

cation traces

Ca: ≤0.005%
Fe: ≤0.002%
K: ≤0.01%
Na: ≤0.05%
Ni: ≤0.01%

greener alternative category

SMILES string

O.CC(=O)O[Cu]OC(C)=O

InChI

1S/2C2H4O2.Cu.H2O/c2*1-2(3)4;;/h2*1H3,(H,3,4);;1H2/q;;+2;/p-2

InChI key

NWFNSTOSIVLCJA-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalysis. Click here for more information.

Application

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rahul Kumar Mishra et al.
Ultrasonics sonochemistry, 56, 55-62 (2019-05-19)
Furfural is a versatile biomass-derived platform compound used for the synthesis of several strategic chemicals. The sonochemically synthesized Zn doped CuO nanoparticles (NPs) were used for the production of furfural. The catalytic activity of the Zn doped CuO NPs was
The kinetics of thermal dehydration of copper (II) acetate monohydrate in air.
Zhang K, et al.
Thermochimica Acta, 437(1), 145-149 (2005)
Dinuclear copper (II) acetate monohydrate: a redetermination of the structure by neutron-diffraction analysis.
Brown GM and Chidambaram R.
Acta Crystallographica Section B, Structural Science, 29(11), 2393-2403 (1973)
F Heidenau et al.
Journal of materials science. Materials in medicine, 16(10), 883-888 (2005-09-17)
Postoperative implant-associated infection is still an unresolved and serious complication in modern surgery. Antibacterial and biocompatible surfaces could both reduce infection rates and promote tissue integration. In this respect, a comparative study of the antibacterial as well as the biocompatible
Amit N Pandya et al.
Tetrahedron letters, 55(50), 6922-6924 (2014-12-17)
An efficient strategy for the synthesis of indolizines from readily available starting materials via oxidative C-H functionalization and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service