Skip to Content
Merck
All Photos(3)

Documents

342688

Sigma-Aldrich

Aluminum oxide

fused, powder, primarily α-phase, -325 mesh

Synonym(s):

Alumina

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al2O3
CAS Number:
Molecular Weight:
101.96
EC Number:
MDL number:
UNSPSC Code:
12352311
eCl@ss:
38120402
PubChem Substance ID:
NACRES:
SB.52

form

fused
powder

particle size

-325 mesh

mp

2040 °C (lit.)

SMILES string

O=[Al]O[Al]=O

InChI

1S/2Al.3O

InChI key

TWNQGVIAIRXVLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of thermal conductivity of SiO2-Al2O3-Y2O3 glasses.
Samal S, et al.
Thermochimica Acta, 604, 1-6 (2015)
Dehydration of 2-butanol by a sulfated zirconia/γ-alumina mixed metal oxide catalyst inside a gas chromatograph injection port.
Skrdla PJ and Lindemann C.
Applied Catalysis A: General, 246(2), 227-235 (2003)
Prescott R and Graham MJ.
Oxidation of Metals, 38(3-4), 233-254 (1992)
Thermoanalytical studies of water on activated alumina, Brockmann I-V,(acid, neutral, basic) from- 60? to+ 700? C.
Hampson JW and Bleam WF.
Thermochimica Acta, 288(1), 179-189 (1996)
Chien-Chih Lin et al.
Nanoscale, 5(17), 8090-8097 (2013-07-25)
We demonstrated a promising route for enhancing temperature sensitivity, improving saturation voltage, and reducing power consumption of the MOS(p) tunneling temperature sensors by introducing ultrathin Al2O3 into the dielectric stacks. Detailed illustrations of the working mechanism and device concept are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service