Skip to Content
Merck
All Photos(1)

Key Documents

267740

Sigma-Aldrich

Aluminum oxide

pore size 58 Å, ~150 mesh

Synonym(s):

Alumina

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al2O3
CAS Number:
Molecular Weight:
101.96
EC Number:
MDL number:
UNSPSC Code:
12352311
eCl@ss:
38120402
PubChem Substance ID:
NACRES:
SB.52

description

weakly acidic

Quality Level

form

powder

technique(s)

ion chromatography: suitable

impurities

<0.03% Fe2O3
<0.4% Na2O

particle size

~150 mesh

pore size

58 Å

pH

~6.0 ( in H2O)

mp

2040 °C (lit.)

anion traces

chloride (Cl-): ~0.06 meq/g
silicate (as SiO2): <0.03%

SMILES string

O=[Al]O[Al]=O

InChI

1S/2Al.3O

InChI key

TWNQGVIAIRXVLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Brockmann aluminum oxides are suitable for column chromatography and adsorptive filtration.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of different catalyst supports on the (n, m) selective growth of single-walled carbon nanotube from Co-Mo catalyst.
Wang B, et al.
J. Mater. Sci., 44(12), 3285-3295 (2009)
Dehydration of 2-butanol by a sulfated zirconia/γ-alumina mixed metal oxide catalyst inside a gas chromatograph injection port.
Skrdla PJ and Lindemann C.
Applied Catalysis A: General, 246(2), 227-235 (2003)
Prescott R and Graham MJ.
Oxidation of Metals, 38(3-4), 233-254 (1992)
Thermoanalytical studies of water on activated alumina, Brockmann I-V,(acid, neutral, basic) from- 60? to+ 700? C.
Hampson JW and Bleam WF.
Thermochimica Acta, 288(1), 179-189 (1996)
Colin J Ingham et al.
Biotechnology advances, 30(5), 1089-1099 (2011-08-23)
Porous aluminum oxide (PAO) is a ceramic formed by an anodization process of pure aluminum that enables the controllable assembly of exceptionally dense and regular nanopores in a planar membrane. As a consequence, PAO has a high porosity, nanopores with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service