Skip to Content
Merck
All Photos(1)

Documents

31434

Sigma-Aldrich

Sodium chloride

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5%

Synonym(s):

Halite, NaCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

vapor pressure

1.33 hPa ( 865 °C)

Assay

≥99.5%

form

solid

impurities

acidity or alkalinity, complies
≤0.0001% hexacyanoferrate(II)
≤0.001% total nitrogen (N)
≤0.005% free acid (as HCl)
≤0.005% free alkali (as NaOH)
≤0.005% in water insoluble matters
≤0.01% Mg, alkaline earth metals (as Ca)

loss

≤0.2% loss on drying, 130 °C

pH

5.0-8.0 (25 °C, 5%)

mp

801 °C (lit.)

solubility

water: soluble 100 mg/mL, clear, colorless

anion traces

bromide (Br-): ≤50 mg/kg
chlorate, nitrate (as NO3-): ≤30 mg/kg
iodide (I-): ≤10 mg/kg
phosphate (PO43-): ≤5 mg/kg
sulfate (SO42-): ≤10 mg/kg

cation traces

Al: ≤0.2 mg/kg
As: ≤0.5 mg/kg
Ba: ≤10 mg/kg
Ca: ≤20 mg/kg
Fe: ≤1 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
heavy metals: ≤3 ppm (by ICP-OES)

suitability

complies for appearance of solution

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Crystals of sodium chloride belong to the cubic crystal system. It is widely employed preservative for meat, vegetable and fish products. Its effect on the growth of salmonellae (mixed inoculum of Salmonella thompson, S. stanley and S. infantis) has been investigated in laboratory medium.

Application

Sodium chloride was used in the dissolution of polymers such as poly(allylamine hydrochloride), sodium poly(styrenesulfonate) and poly(acrylic acid).

Packaging

This product is available in HdPe bottles.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Zander et al.
Molecules (Basel, Switzerland), 25(17) (2020-09-02)
Hyaluronan is an essential physiological bio macromolecule with different functions. One prominent area is the synovial fluid which exhibits remarkable lubrication properties. However, the synovial fluid is a multi-component system where different macromolecules interact in a synergetic fashion. Within this
Stefaan Derveaux et al.
Analytical chemistry, 80(1), 85-94 (2007-12-07)
The "layer-by-layer" (LbL) technology has been widely investigated for the coating of flat substrates and capsules with polyelectrolytes. In this study, LbL polyelectrolyte coatings applied at the surface of digitally encoded microcarriers were evaluated for the quantitative, sensitive, and simultaneous
Xi Chen et al.
Current biology : CB, 30(8), 1477-1490 (2020-03-21)
How septin architecture is remodeled from an hourglass to a double ring during cytokinesis in fungal and animal cells remains unknown. Here, we show that during the hourglass-to-double-ring transition in budding yeast, septins acquire a "zonal architecture" in which paired
Ruitao Su et al.
Science advances, 6(41) (2020-10-11)
Microfluidic devices fabricated via soft lithography have demonstrated compelling applications such as lab-on-a-chip diagnostics, DNA microarrays, and cell-based assays. These technologies could be further developed by directly integrating microfluidics with electronic sensors and curvilinear substrates as well as improved automation
Shalini Iyer et al.
PeerJ, 6, e5815-e5815 (2018-10-26)
Amyotrophic lateral sclerosis (ALS) and frontotemporal dementia (FTD), two late onset neurodegenerative diseases, have been shown to share overlapping cellular pathologies and genetic origins. Studies suggest that a hexanucleotide repeat expansion in the first intron of the C9orf72 gene is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service