Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Calcium oxide
Synonym(s):
Lime, Quicklime
Linear Formula:
CaO
CAS Number:
1305-78-8
Molecular Weight:
56.08
EC Number:
215-138-9
Products
Product Category
salts (1)
Brand
Sigma-Aldrich (5)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Reagent Type
Core
Special Grade
Anhydrous (1)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
248568
Calcium oxide,
reagent grade
Expand
View Pricing
208159
Calcium oxide,
ReagentPlus
®
, 99.9% trace metals basis
Expand
View Pricing
451711
Calcium oxide,
anhydrous, powder, ≥99.99% trace metals basis
Expand
View Pricing
634182
Calcium oxide,
nanopowder, <160 nm particle size (BET), 98%
Expand
View Pricing
229539
Calcium oxide,
99.995% trace metals basis
Expand
View Pricing