Skip to Content
Merck
All Photos(3)

Key Documents

221279

Sigma-Aldrich

Manganese(II) chloride tetrahydrate

ACS reagent, ≥98%

Synonym(s):

Manganous chloride tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MnCl2 · 4H2O
CAS Number:
Molecular Weight:
197.91
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent

Quality Level

Assay

≥98%
98.0-101.0% (ACS specification)

form

crystals

impurities

≤0.005% insolubles

pH

3.5-6.0 (25 °C, 5%)

mp

58 °C (lit.)

anion traces

sulfate (SO42-): ≤0.005%

cation traces

Ca: ≤0.005%
Fe: ≤5 ppm
K: ≤0.01%
Mg: ≤0.005%
Na: ≤0.05%
Zn: ≤0.005%
heavy metals: ≤5 ppm (by ICP)

SMILES string

Cl[Mn]Cl.[H]O[H].[H]O[H].[H]O[H].[H]O[H]

InChI

1S/2ClH.Mn.4H2O/h2*1H;;4*1H2/q;;+2;;;;/p-2

InChI key

CNFDGXZLMLFIJV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Manganese(II) chloride tetrahydrate may be used to accelerate the synthesis of dihydropyrimidin-2(1H)-ones by the condensation of aldehyde, β-dicarbonyl compounds and urea under microwave conditions. It may also be used in the synthesis of manganese(II) tris(2-benzimidazolylmethyl)amine (ntb) complexes.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - STOT RE 2

Target Organs

Brain

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vahid Safarifard et al.
Ultrasonics sonochemistry, 21(1), 253-261 (2013-06-22)
Nanoparticles of a new three-dimensional Mn(II) coordination supramolecular compound, [Mn(L)2(H2O)2] (1), (L(-)=1H-1,2,4-triazole-3-carboxylate), have been synthesized by a sonochemical process and characterized by scanning electron microscopy (SEM), X-ray powder diffraction (XRPD), FT-IR spectroscopy and elemental analyses. Structural determination of compound 1
Myoung Soo Lah et al.
Inorganic chemistry, 36(9), 1782-1785 (1997-04-23)
A series of manganese(II) tris(2-benzimidazolylmethyl)amine (ntb) complexes were synthesized and characterized. In these complexes, ntb functions as a tripodal tetradentate ligand, resulting in trigonal pyramidal geometry. The complexes have five- to seven-coordinate manganese(II) ions depending on the additional ligand used.
D J Sánchez et al.
Toxicology letters, 69(1), 45-52 (1993-07-01)
Manganese (II) chloride tetrahydrate was investigated in Swiss mice for maternal and developmental toxicity after subcutaneous (s.c.) exposure to doses of 0, 2, 4, 8 and 16 mg/kg per day from gestation day 6 through 15. Females were sacrificed on
Cobalt (II) chloride or manganese (II) chloride or tin (II) chloride promoted one pot synthesis of dihydropyrimidin-2 (1H)-ones using microwave irradiation.
Kumar S, et al.
Indian Journal of Chemistry, 44B, 762-767 (2005)
Adam B Robertson et al.
Nature protocols, 7(2), 340-350 (2012-01-28)
We describe a method for the efficient and selective identification of DNA containing the 5-hydroxymethylcytosine (5-hmC) modification. This protocol takes advantage of two proteins: T4 β-glucosyltransferase (β-gt), which converts 5-hmC to β-glucosyl-5-hmC (β-glu-5-hmC), and J-binding protein 1 (JBP1), which specifically

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service