Skip to Content
Merck
All Photos(2)

Key Documents

C8661

Sigma-Aldrich

Cobalt(II) chloride hexahydrate

BioReagent, suitable for cell culture, suitable for insect cell culture

Synonym(s):

Cobaltous chloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CoCl2 · 6H2O
CAS Number:
Molecular Weight:
237.93
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.71
grade:
reagent
form:
powder

grade

reagent

vapor pressure

40 mmHg ( 0 °C)

product line

BioReagent

form

powder

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable

SMILES string

O.O.O.O.O.O.Cl[Co]Cl

InChI

1S/2ClH.Co.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

GFHNAMRJFCEERV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

  • Synthesis, Characterization, and Biological Activities of Novel Vanadium(IV) and Cobalt(II) Complexes: Investigates the synthesis of novel cobalt(II) complexes and their potential biological applications, particularly emphasizing their bioactive properties in various cellular models (Damena T, Zeleke D, Desalegn T, et al. 2022. ACS Omega).

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Dam. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Baoan Hong et al.
International journal of biological sciences, 15(3), 587-597 (2019-02-13)
Tribbles pseudokinase 3 (TRIB3) is a member of the mammalian pseudokinase tribbles family and is involved in multiple biological processes. However, the role of TRIB3 in renal cell carcinoma (RCC) remains unclear. In this study, we aimed to elucidate the
Shahna Shahulhameed et al.
Frontiers in neuroscience, 13, 1445-1445 (2020-03-03)
The detailed mechanisms underlying oxidative stress that leads to neuroinflammation and neurodegeneration in retinal vascular conditions, including diabetic retinopathy, retinopathy of prematurity etc., remain largely unexplored mainly due to a lack of suitable disease models that can simulate the inherent
Tugba Kilic et al.
Scientific reports, 8(1), 9402-9402 (2018-06-22)
Nanoscale extracellular vesicles (EVs) including exosomes (50-150 nm membrane particles) have emerged as promising cancer biomarkers due to the carried genetic information about the parental cells. However the sensitive detection of these vesicles remains a challenge. Here we present a label-free
Yuan Zhang et al.
Clinica chimica acta; international journal of clinical chemistry, 489, 203-211 (2017-12-11)
Accumulated data indicate that placental hypoxia is implicated in the pathogenesis of preeclampsia (PE). Tight junction (TJ) is important structure that sustains normal placental barrier function, its dysregulation under hypoxia has been observed. This study was designed to explore hypoxia-induced
Seo-Hyun Choi et al.
Biochimica et biophysica acta. Gene regulatory mechanisms, 1862(8), 771-785 (2019-07-05)
Overexpressed Solute Carrier Family 16 Member 3 (SLC16A3, also called MCT4) plays a critical role in hypoxic cancer cell growth and proliferation, by expelling glycolysis-derived lactate across the plasma membrane. However, how SLC16A3 expression is regulated, under hypoxic conditions, is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service