All Photos(1)
About This Item
Linear Formula:
HBF4
CAS Number:
Molecular Weight:
87.81
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21
Recommended Products
vapor density
3 (vs air)
Quality Level
vapor pressure
5 mmHg ( 20 °C)
form
liquid
concentration
46.0-52.0% in NaOH (titration)
48 wt. % in H2O
density
1.4 g/mL at 25 °C
SMILES string
F.FB(F)F
InChI
1S/BF3.FH/c2-1(3)4;/h;1H
InChI key
LEMQFBIYMVUIIG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Tetrafluoroboric acid is used as a catalyst for the protection and deprotection reactions of various carbohydrates. It participates in the synthesis of 4-sulfonic acid phenyl diazonium tetrafluoroborate, which was required for the preparation of sulfonated graphene (SG).
Application
Tetrafluoroboric acid solution may be used as a catalyst for the hydration of aromatic haloalkynes to α-halomethyl ketones in the absence of metal catalysts. It may also be used the epoxidized soybean oil (ESBO) ring opening step of fatty acids preparation.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Tetrafluoroboric acid, an efficient catalyst in carbohydrate protection and deprotection reactions.
Albert R, et al.
Carbohydrate Research, 137, 282-290 (1985)
Polyols and polyurethanes prepared from epoxidized soybean oil ring-opened by polyhydroxy fatty acids with varying OH numbers.
Chen R, et al.
Journal of Applied Polymer Science, 132(1) (2015)
Metal-free hydration of aromatic haloalkynes to a-halomethyl ketones.
Ye M, et al.
Tetrahedron Letters, 57(45), 4983-4986 (2016)
M P Dariel et al.
Dental materials : official publication of the Academy of Dental Materials, 11(3), 208-217 (1995-05-01)
The purpose of this study was to develop a new technology for preparing mercury-free metallic dental restorative materials. The novel approach relies on the cold welding of surface treated silver particles. At ambient temperature, intermetallic compound formation takes place spontaneously
Dong-Su Kim et al.
Organic letters, 12(5), 1092-1095 (2010-02-13)
A novel series of alkyne-containing potassium organotrifluoroborates were prepared in good yields from the corresponding haloaryltrifluoroborates and various alkynes via Sonogashira coupling reaction. Also, the Suzuki-Miyaura cross-coupling reaction of alkynylaryltrifluoroborates with aryl and alkenyl bromides was achieved in the presence
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service