Skip to Content
Merck
All Photos(3)

Key Documents

14027

Sigma-Aldrich

Titanium(IV) oxide

puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5%

Synonym(s):

Titania, Titanium dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiO2
CAS Number:
Molecular Weight:
79.87
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
form:
powder
Pricing and availability is not currently available.

grade

puriss.

Quality Level

Assay

98.0-100.5%
99-100.5%
99.0-100.5% (calc. on dry substance)

form

powder

quality

meets analytical specification of Ph. Eur., BP, USP

impurities

acidity or alkalinity, complies
residual solvents, complies
≤0.002% heavy metals (as Pb)
≤0.25% water-soluble matter
≤0.5% in acid (HCl 0,5 mol/l) sol. matter
≤0.5% water-soluble matter (hot)

loss

≤0.5% loss on drying, 105 °C, 3h
≤0.5% loss on ignition, 800 °C

mp

>350 °C (lit.)

density

4.26 g/mL at 25 °C (lit.)

cation traces

As: ≤1 mg/kg
Ba:, in accordance
Fe: ≤50 mg/kg
Pb: ≤20 mg/kg
Sb: ≤50 mg/kg

suitability

complies for appearance of solution

SMILES string

O=[Ti]=O

InChI

1S/2O.Ti

InChI key

GWEVSGVZZGPLCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Titanium dioxide (TiO2, titania) is one of the most important materials having various important applications, such as water and air purification, self-cleaning materials and photovoltaic cells. TiO2 is an n-type semiconductor having a wide band gap (3.2eV for anatase and 3.0eV for rutile). Deposition of thin films of TiO2 doped with Mn on F-doped SnO2-coated glass by spin coating has been described. Deposition of thin films of titanium dioxide (TiO2) on various substrates by a simple sol-gel dip coating technique has been proposed.[1] It has various photocatalytic applications where it can be used in two forms, i.e as highly dispersed fine particles on porous support materials and as suspended fluids in liquid medium.[2]
Titanium(IV) oxide is a white colored compound, which can be used as a coloring pigment[3] in paints and a main ingredient of cosmetics and toothpaste.[4] It can be prepared via reaction of titanium(IV) chloride with oxygen gas.[3] It can also find applications in photovoltaics, photocatalysis and gas sensors.[5]

Application


  • Structure, properties, production and photocatalytic processes of titanium (IV) oxide.: This study discusses the structure, properties, and production methods of titanium(IV) oxide, highlighting its photocatalytic processes which are essential in environmental applications such as pollutant degradation (Byts et al., 2023).

  • Photocatalytic Activity of Radial Rutile Titanium(IV) Oxide Microspheres for Aerobic Oxidation of Organics.: This research investigates the photocatalytic performance of radial rutile TiO2 microspheres, modified with iron oxide clusters, in the aerobic oxidation of organic compounds, demonstrating their potential in environmental cleanup (Kojima et al., 2022).

  • Photocatalytic degradation of textile dye with titanium (IV) doped tungsten oxide nanoparticles.: This study focuses on the photocatalytic degradation of textile dyes using Ti(IV)-doped tungsten oxide nanoparticles under visible light, highlighting their potential in water purification due to enhanced photocatalytic activity (Ranjin et al., 2024).

  • Solubility and Surface Thermodynamics of Conducting Polymers by Inverse Gas Chromatography, IV: Polypyrrole/Titanium Oxide.: This paper examines the solubility and surface thermodynamics of polypyrrole/titanium oxide composites using inverse gas chromatography, providing insights into their application in electronic materials (Ballard & Al-Saigh, 2020).

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of a titanium dioxide-supported platinum catalyst with ultrahigh stability for polymer electrolyte membrane fuel cell applications
Huang S-Y, et al.
Journal of the American Chemical Society, 131(39), 13898-13899 (2009)
Zumdahl.SS and DeCoste.JD
Introductory Chemistry, (7) (2010)
Defect chemistry of titanium dioxide. Application of defect engineering in processing of TiO2-based photocatalysts
Nowotny.K.M, et al.
The Journal of Physical Chemistry C, 112(14), 5275-5300 (2008)
Preparation of titanium (IV) oxide thin film photocatalyst by sol-gel dip coating.
Sonawane RS, et al.
Materials Chemistry and Physics, 77(3), 744-750 (2003)
Multivalent Mn-doped TiO2 thin films.
Lin CYW, et al.
Physica E: Low-Dimensional Systems and Nanostructures, 44(!0), 1969-1972 (2012)

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Is the TiO2 in rutile or anatase form in this specific product? And is this a micron powder?

    1 answer
    1. The crystalline structure is anatase. The d50 particle size for this product has historically been between 2 and 3 microns, but that is an approximation and particle size information does not appear on the COA for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service