Skip to Content
Merck
All Photos(1)

Key Documents

Y0001236

Econazole

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H15Cl3N2O
CAS Number:
Molecular Weight:
381.68
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

econazole

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Clc1c(ccc(c1)Cl)C(OCc3ccc(cc3)Cl)C[n]2cncc2

InChI

1S/C18H15Cl3N2O/c19-14-3-1-13(2-4-14)11-24-18(10-23-8-7-22-12-23)16-6-5-15(20)9-17(16)21/h1-9,12,18H,10-11H2

InChI key

LEZWWPYKPKIXLL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Econazole EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott Thomson et al.
Scientific reports, 7(1), 8247-8247 (2017-08-16)
The soil amoebae Acanthamoeba causes Acanthamoeba keratitis, a severe sight-threatening infection of the eye and the almost universally fatal granulomatous amoebic encephalitis. More effective treatments are required. Sterol biosynthesis has been effectively targeted in numerous fungi using azole compounds that
Chao Dong et al.
Scientific reports, 7(1), 17987-17987 (2017-12-23)
The phosphatidylinositol-3-kinase (PI3K)/AKT signaling pathway plays a pivotal role in many cellular processes, including the proliferation, survival and differentiation of lung cancer cells. Thus, PI3K is a promising therapeutic target for lung cancer treatment. In this study, we applied free
Wei Qiu et al.
Archives of oral biology, 73, 113-120 (2016-10-21)
The aim of this study was to determine the inhibitory effect of eight antifungal drugs on S. mutans growth, biofilm formation and virulence factors. The actions of antifungal drugs on S. mutans were determined by recovery plates and survival kinetic
Xianzhi Peng et al.
The Science of the total environment, 426, 311-317 (2012-04-24)
Residue of azole antifungals in the environment is of concern due to the environmental risks and persistence. Distribution, behavior, and fate of frequently used azole antifungal pharmaceuticals were investigated in wastewater at two sewage treatment plants (STPs) in China. Fluconazole
Azza A Mahmoud et al.
International journal of pharmaceutics, 413(1-2), 229-236 (2011-05-05)
Development of efficient ocular delivery nanosystems remains a major challenge to achieve sustained therapeutic effect. The purpose of this work was to develop chitosan nanoparticles using sulfobutylether-β-cyclodextrin (SBE-β-CD) as polyanionic crosslinker and to investigate the potential of using those nanostructures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service