Skip to Content
Merck
All Photos(1)

Documents

E4632

Sigma-Aldrich

Econazole nitrate salt

Synonym(s):

1-(2-[(4-Chlorophenyl)methoxy]-2-[2,4-dichlorophenyl]ethyl)-1H-imidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H15Cl3N2O · HNO3
CAS Number:
Molecular Weight:
444.70
EC Number:
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85

form

powder or crystals

Quality Level

color

white to off-white

antibiotic activity spectrum

Gram-positive bacteria
fungi

Mode of action

cell membrane | interferes
cell wall synthesis | interferes
enzyme | inhibits

SMILES string

O[N+]([O-])=O.Clc1ccc(COC(Cn2ccnc2)c3ccc(Cl)cc3Cl)cc1

InChI

1S/C18H15Cl3N2O.HNO3/c19-14-3-1-13(2-4-14)11-24-18(10-23-8-7-22-12-23)16-6-5-15(20)9-17(16)21;2-1(3)4/h1-9,12,18H,10-11H2;(H,2,3,4)

InChI key

DDXORDQKGIZAME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: imidazole

Application

Econazole is a broad spectrum antimycotic similar to ketoconazole. It has some action against Gram positive bacteria. It is used topically in dermatomycoses as well as orally and parenterally. It is used for studies on processes such as platelet serotonin uptake, prostanoid biosynthesis, EDHF-mediated relaxation, and Ca2+ transport pathways in thymic lymphocytes.

Biochem/physiol Actions

Econazole interacts with 14-α demethylase, a cytochrome P-450 enzyme necessary to convert lanosterol to ergosterol. Inhibition of ergosterol results in increased cellular permeability causing leakage of cellular contents. Econazole may also inhibit endogenous respiration, interact with membrane phospholipids, inhibit the transformation of yeasts to mycelial forms, inhibit purine uptake, and impair triglyceride and phospholipid biosynthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Núria Daranas et al.
PloS one, 13(1), e0190931-e0190931 (2018-01-06)
Lactobacillus plantarum strains PM411 and TC92 can efficiently control bacterial plant diseases, but their fitness on the plant surface is limited under unfavourable low relative humidity (RH) conditions. To increase tolerance of these strains to water stress, an adaptive strategy
Núria Daranas et al.
Applied and environmental microbiology, 84(10) (2018-03-11)
A viability quantitative PCR (v-qPCR) assay was developed for the unambiguous detection and quantification of Lactobacillus plantarum PM411 viable cells in aerial plant surfaces. A 972-bp region of a PM411 predicted prophage with mosaic architecture enabled the identification of a
C J Jackson et al.
FEMS microbiology letters, 192(2), 159-162 (2000-11-07)
Azole antifungals are central to therapy and act by inhibiting a cytochrome P450, sterol 14-demethylase and blocking normal sterol synthesis. Our recent identification of a mycobacterial sterol biosynthetic pathway led us to probe the efficacy of a range of these
Inhibition of Ca2+ transport pathways in thymic lymphocytes by econazole, miconazole, and SKF 96365
M. J. Mason, B. Mayer, et al.
American Journal of Physiology. Cell Physiology, 264, C654-C662 (1993)
M A Ghannoum et al.
Clinical microbiology reviews, 12(4), 501-517 (1999-10-09)
The increased use of antibacterial and antifungal agents in recent years has resulted in the development of resistance to these drugs. The significant clinical implication of resistance has led to heightened interest in the study of antimicrobial resistance from different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service