Skip to Content
Merck
All Photos(1)

Key Documents

Y0001141

Diltiazem for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(+)-cis-Diltiazem hydrochloride, (2S,3S)-(+)-cis-3-Acetoxy-5-(2-dimethylaminoethyl)-2,3-dihydro-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one hydrochloride, CRD-401

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26N2O4S · HCl
CAS Number:
Molecular Weight:
450.98
Beilstein:
4228706
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

diltiazem

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.COc1ccc(cc1)[C@@H]2Sc3ccccc3N(CCN(C)C)C(=O)[C@@H]2OC(C)=O

InChI

1S/C22H26N2O4S.ClH/c1-15(25)28-20-21(16-9-11-17(27-4)12-10-16)29-19-8-6-5-7-18(19)24(22(20)26)14-13-23(2)3;/h5-12,20-21H,13-14H2,1-4H3;1H/t20-,21+;/m1./s1

InChI key

HDRXZJPWHTXQRI-BHDTVMLSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Diltiazem for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mustafa Kayan et al.
The Journal of membrane biology, 245(12), 833-840 (2012-08-21)
Non-ionic contrast media (CM) can induce tissue kidney injury via activation of phagocytosis and oxidative stress, although the mechanisms of injury via neutrophils are not clear. We investigated the effects of CM on oxidative stress and Ca²⁺ concentrations in serum
Diltiazem hydrochloride.
B F McGraw
Drug intelligence & clinical pharmacy, 16(5), 366-370 (1982-05-01)
Joanna E Barbara et al.
Drug metabolism and disposition: the biological fate of chemicals, 40(10), 1966-1975 (2012-07-17)
In vitro metabolite profiling and characterization experiments are widely employed in early drug development to support safety studies. Samples from incubations of investigational drugs with liver microsomes or hepatocytes are commonly analyzed by liquid chromatography/mass spectrometry for detection and structural
Toshio Obata et al.
Clinical biochemistry, 46(1-2), 164-166 (2012-09-25)
The interaction between the PKC and oxygen free radicals. The present study examined the effects of lysophosphatidylcholine (LPC), an endogenous amphophilic lipid metabolite, on 1-methyl-4-phenylpyridinium ion (MPP(+))-induced hydroxyl radical (()OH) generation in rat striatum. LPC (50 μM) increased ()OH formation
[Diltiazem hydrochloride: synthetic and pharmacological studies and development].
K Abe et al.
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 108(8), 716-732 (1988-08-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service