Skip to Content
Merck
All Photos(1)

Key Documents

Y0000165

Tribenoside

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Ethyl 3,5,6-tri-O-benzyl-D-glucofuranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H34O6
CAS Number:
Molecular Weight:
478.58
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

tribenoside

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

shipped in

wet ice

storage temp.

−20°C

SMILES string

O1[C@@H]([C@@H]([C@H](C1OCC)O)OCc4ccccc4)C(OCc3ccccc3)COCc2ccccc2

InChI

1S/C29H34O6/c1-2-32-29-26(30)28(34-20-24-16-10-5-11-17-24)27(35-29)25(33-19-23-14-8-4-9-15-23)21-31-18-22-12-6-3-7-13-22/h3-17,25-30H,2,18-21H2,1H3/t25?,26-,27-,28-,29?/m1/s1

InChI key

ULLNJSBQMBKOJH-NUZMXOBKSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tribenoside EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Chayen et al.
Cell biochemistry and function, 14(1), 57-61 (1996-03-01)
Up to nine out of 10 male STR/ORT mice develop osteoarthritis (OA) of the medial tibial cartilage at an early age. This has now been shown to be related to changes in the activity and distribution of monoamine oxidase which
A Sioufi et al.
European journal of drug metabolism and pharmacokinetics, 7(3), 223-228 (1982-01-01)
In an experiment on three healthy volunteers, plasma levels of tribenoside and the elimination of benzoic acid in plasma and urine were followed up during a 7-day period of daily oral medication with tribenoside. Plasma levels of tribenoside, and plasma
M Horváth et al.
Allergologia et immunopathologia, 10(1), 25-32 (1982-01-01)
The in vitro effects of azathioprine, clofibrate and tribenoside on the cell-mediated immune response was studied in some vascular diseases. Imuran decreased leucocyte migration inhibition induced by vascular antigens in patients with myocardial infarction, peripheral arterial disease or superficial thrombophlebitis.
[Clinical experiences with a vasoprotective drug in the treatment of venous problems of the lower limbs].
M De Caprio et al.
La Clinica terapeutica, 101(5), 497-508 (1982-06-15)
Glyvenol therapy in varicose veins an evaluation of its effectiveness.
I Kakande
East African medical journal, 58(7), 532-538 (1981-07-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service