Skip to Content
Merck
All Photos(1)

Key Documents

Y0001082

Fluvastatin for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Fluvastatin sodium, (3R,5S,6E)-rel-7-[3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3,5-dihydroxy-6-heptenoic acid monosodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H25FNNaO4
CAS Number:
Molecular Weight:
433.45
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

fluvastatin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Na+].CC(C)n1c(\C=C\[C@H](O)C[C@H](O)CC([O-])=O)c(-c2ccc(F)cc2)c3ccccc13

InChI

1S/C24H26FNO4.Na/c1-15(2)26-21-6-4-3-5-20(21)24(16-7-9-17(25)10-8-16)22(26)12-11-18(27)13-19(28)14-23(29)30;/h3-12,15,18-19,27-28H,13-14H2,1-2H3,(H,29,30);/q;+1/p-1/b12-11+;/t18-,19-;/m0./s1

InChI key

ZGGHKIMDNBDHJB-RPQBTBOMSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fluvastatin for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allen B Williams et al.
Blood, 120(15), 3069-3079 (2012-08-29)
FLT3 is frequently mutated in acute myeloid leukemia (AML), but resistance has limited the benefit of tyrosine kinase inhibitors (TKI). We demonstrate that statins can impair FLT3 glycosylation, thus leading to loss of surface expression and induction of cell death
C Kondo et al.
Journal of viral hepatitis, 19(9), 615-622 (2012-08-07)
Pegylated interferon (PEG-IFN)/ribavirin combination therapy is the standard-of-care (SOC) treatment for chronic hepatitis C patients infected with hepatitis C virus (HCV) genotype 1b and high viral load. The addition of fluvastatin to SOC treatment has been suggested to be effective
T Bader et al.
Journal of viral hepatitis, 20(9), 622-627 (2013-08-06)
Fluvastatin or simvastatin has demonstrable antiviral activity against hepatitis C virus (HCV) as monotherapy. The safety and efficacy of adding fluvastatin or simvastatin to peginterferon/ribavirin for 48 weeks was tested in HCV genotype 1 naïve-to-treatment veterans. Thirty-seven naïve-to-treatment genotype 1
Masanori Atsukawa et al.
Journal of gastroenterology and hepatology, 28(1), 51-56 (2012-09-20)
Although the anti-hepatitis C virus (HCV) effect of statins in vitro and clinical efficacy of fluvastatin combined with Pegylated interferon (PEG-IFN)/ribavirin therapy for chronic hepatitis C (CHC) have been reported, the details of clinical presentation are largely unknown. We focused
Janine Kah et al.
Oncology reports, 28(3), 1077-1083 (2012-06-20)
HMG-CoA-reductase inhibitors (statins) are widely used drugs to interfere with cholesterol biosynthesis. Besides this usage, evidence is increasing that statins might also be useful in therapy of viral infections or cancer. We investigated the effects of fluva-, simva-, atorva-, rosuva-

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service