Skip to Content
Merck
All Photos(1)

Documents

PHR1620

Supelco

Fluvastatin sodium

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

(3R,5S,6E)-rel-7-[3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3,5-dihydroxy-6-heptenoic acid monosodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H25FNNaO4
CAS Number:
Molecular Weight:
433.45
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0001082
traceable to USP 1285931

API family

fluvastatin

CofA

current certificate can be downloaded

packaging

ampule of 1 × 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

[Na+].CC(C)n1c(\C=C\[C@H](O)C[C@H](O)CC([O-])=O)c(-c2ccc(F)cc2)c3ccccc13

InChI

1S/C24H26FNO4.Na/c1-15(2)26-21-6-4-3-5-20(21)24(16-7-9-17(25)10-8-16)22(26)12-11-18(27)13-19(28)14-23(29)30;/h3-12,15,18-19,27-28H,13-14H2,1-2H3,(H,29,30);/q;+1/p-1/b12-11+;/t18-,19-;/m0./s1

InChI key

ZGGHKIMDNBDHJB-RPQBTBOMSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Fluvastatin sodium is a lipid-lowering drug that belongs to the group of compounds known as statins. It acts by inhibiting the enzyme HMG-CoA reductase that plays a key role in the cholesterol production. It is generally used to reduce plasma cholesterol levels and prevent cardiovascular disease.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Fluvastatin sodium may be used as a reference standard for the determination of the analyte in bulk drug and pharmaceutical formulations by high performance liquid chromatography.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA5290 in the slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

The role of statin drugs in combating cardiovascular diseases-A review
Pichandi S, et al
International journal of current science research, 1(2), 47-56 (2011)
Fluvastatin Sodium
USP42/NF37: United States Pharmacopeia and National Formulary
United States Pharmacopeia/National Formulary, 43(5), 1972-1972 (2019)
Development and validation of stability-indicating HPLC methods for quantitative determination of pravastatin, fluvastatin, atorvastatin, and rosuvastatin in pharmaceuticals
Gomes FP, et al.
Analytical Letters, 42(12), 1784-1804 (2009)
Determination of Fluvastatin Sodium Extended-release Tablets by HPLC
Zhang S, et al.
China Pharmacy, 42(34), 1784-1804 (2007)
Masao Nishiya et al.
Anti-cancer drugs, 30(5), 451-457 (2019-03-29)
Although vemurafenib has been shown to improve the overall survival of patients with metastatic melanoma harboring the BRAF V600E mutation, its efficacy is often hampered by drug resistance acquired within a relatively short period through several distinct mechanisms. In the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service