Skip to Content
Merck
All Photos(1)

Key Documents

Y0001075

Rifaximin for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Rifaximin, 4-Deoxy-4′-methylpyrido[1′,2′-1,2]imidazo[5,4-c]rifamycin SV, Rifacol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H51N3O11
CAS Number:
Molecular Weight:
785.88
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

rifaximin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CO[C@H]1\C=C\O[C@@]2(C)Oc3c(C)c(O)c4c(O)c(NC(=O)C(C)=C\C=C\[C@@H](C)[C@@H](O)[C@@H](C)[C@H](O)[C@H](C)[C@H]([C@H]1C)C(=O)OC)c5c(nc6cc(C)ccn56)c4c3C2=O

InChI

1S/C43H51N3O11/c1-19-14-16-46-27(18-19)44-32-29-30-37(49)25(7)39-31(29)40(51)43(8,57-39)56-17-15-26(54-9)22(4)28(42(53)55-10)23(5)36(48)24(6)35(47)20(2)12-11-13-21(3)41(52)45-33(34(32)46)38(30)50/h11-18,20,22-24,26,28,35-36,47-50H,1-10H3,(H,45,52)/b12-11+,17-15+,21-13-/t20-,22+,23-,24-,26+,28+,35-,36-,43+/m1/s1

InChI key

HIYLTQREEOINNF-HTEWPBCCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Rifaximin for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenneth R Lawrence et al.
Pharmacotherapy, 28(8), 1019-1032 (2008-07-29)
To review the effectiveness and safety of rifaximin for the treatment of hepatic encephalopathy. A literature search was conducted of MEDLINE (1966-September 2007), the Cochrane Database of Systematic Reviews (1995-2007), and the Cochrane Hepato-Biliary Group Reviews (2003-2007). English-language articles identified
A Moretti et al.
La Clinica terapeutica, 163(1), 33-38 (2012-03-01)
Diverticular disease of the colon is the fifth most important gastrointestinal disease in terms of direct and indirect healthcare costs in western countries. Although most patients with colonic diverticula remain asymptomatic for their whole life, in 20-25% of cases will
Hoonmo L Koo et al.
Current opinion in gastroenterology, 26(1), 17-25 (2009-11-03)
Rifaximin is gaining attention for its potential activity in a multitude of gastrointestinal diseases. We review the unique pharmaceutical properties of this antibiotic and the published evidence in the literature regarding the use of rifaximin for different gastrointestinal disorders. Rifaximin
Ron Schey et al.
Expert review of gastroenterology & hepatology, 5(4), 461-464 (2011-07-26)
Alterations in gut flora may play an important role in the pathophysiology of bowel symptoms, especially in patients with irritable bowel syndrome (IBS). If so, antibiotics that affect gut flora may offer a novel approach for the management of patients
Filippo Cremonini et al.
Expert opinion on pharmacotherapy, 13(3), 433-440 (2012-01-19)
Few therapeutic options are available for irritable bowel syndrome (IBS). Lubiprostone is approved by the FDA for IBS with constipation, and alosetron in IBS with diarrhea (IBS-D). It has been proposed that alterations in the bowel microflora may play a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service