Skip to Content
Merck
All Photos(1)

Key Documents

Y0001046

Sevoflurane

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1,1,1,3,3,3-Hexafluoro-2-(fluoromethoxy)propane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3F7O
CAS Number:
Molecular Weight:
200.05
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sevoflurane

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

FC(F)(F)C(OCF)C(F)(F)F

InChI

1S/C4H3F7O/c5-1-12-2(3(6,7)8)4(9,10)11/h2H,1H2

InChI key

DFEYYRMXOJXZRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sevoflurane EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maj-Lis Lindholm et al.
Anesthesia and analgesia, 118(4), 782-787 (2014-03-22)
Preclinical data indicate that anesthesia and surgery may promote cancer growth. We previously found no increased risk of malignant disease within 5 years regarding duration of general anesthesia (TANESTH) and time with Bispectral Index (BIS) under 45 (TBIS < 45)
Jan Kehler et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 55(9), 1513-1518 (2014-07-06)
Phosphodiesterase 10A (PDE10A) plays a key role in the regulation of brain striatal signaling, and several pharmaceutical companies currently investigate PDE10A inhibitors in clinical trials for various central nervous system diseases. A PDE10A PET ligand may provide evidence that a
Norifumi Kuratani et al.
Anesthesiology, 109(2), 225-232 (2008-07-24)
Sevoflurane is a popular inhalational anesthetic for general anesthesia in children. The higher incidence of emergence agitation has been suspected after sevoflurane anesthesia as compared with halothane, whereas some controlled studies showed conflicting results. In this report, the authors performed
G Landoni et al.
British journal of anaesthesia, 111(6), 886-896 (2013-07-16)
Many studies have compared desflurane, isoflurane, sevoflurane, total i.v. anaesthesia (TIVA), or all in cardiac surgery to assess their effects on patient survival. We performed standard pairwise and Bayesian network meta-analyses; the latter allows indirect assessments if any of the
Joachim Erb et al.
The Journal of international medical research, 42(3), 750-764 (2014-05-02)
Levosimendan is an inotropic drug with organ-protective properties due to its activation of mitochondrial K(ATP) channels. This prospective, randomized, double-blind, placebo-controlled study investigated whether administration of levosimendan prior to cardiopulmonary bypass could reduce organ dysfunction and influence subsequent secondary endpoints.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service