Skip to Content

1171900

USP

Desflurane

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-(Difluoromethoxy)-1,1,1,2-tetrafluoroethane, Difluoromethyl 1,2,2,2-tetrafluoroethyl ether

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C3H2F6O
CAS Number:
Molecular Weight:
168.04
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

desflurane

manufacturer/tradename

USP

application(s)

pharmaceutical
pharmaceutical (small molecule)

format

neat

SMILES string

FC(F)(F)C(F)OC(F)F

InChI

1S/C3H2F6O/c4-1(3(7,8)9)10-2(5)6/h1-2H

InChI key

DPYMFVXJLLWWEU-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Desflurane USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Eye Irrit. 2 - Repr. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class

10 - Combustible liquids

Flash Point (°F)

>200.0 °F

Flash Point (°C)

> 93.33 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library