Skip to Content
Merck
All Photos(1)

Key Documents

Y0001022

Iopromide for system suitability 2

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Iopromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24I3N3O8
CAS Number:
Molecular Weight:
791.11
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

iopromide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C(N(C)CC(O)CO)C1=C(I)C(NC(COC)=O)=C(I)C(C(NCC(CO)O)=O)=C1I

InChI

1S/C18H24I3N3O8/c1-24(4-9(28)6-26)18(31)12-13(19)11(17(30)22-3-8(27)5-25)14(20)16(15(12)21)23-10(29)7-32-2/h8-9,25-28H,3-7H2,1-2H3,(H,22,30)(H,23,29)

InChI key

DGAIEPBNLOQYER-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Iopromide for system suitability 2 EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arthur Nasis et al.
Radiology, 272(3), 674-682 (2014-04-18)
To assess the long-term outcome and hospital readmission rate associated with a computed tomographic (CT) angiography-guided strategy used to examine patients who present to the emergency department (ED) with symptoms of possible acute coronary syndrome (ACS). The study was approved
Ralph Gnannt et al.
Investigative radiology, 50(1), 17-23 (2014-09-10)
Mortality from abdominal abscesses ranges from 30% in treated cases up to 80% to 100% in patients with undrained or nonoperated abscesses. Various computed tomographic (CT) imaging features have been suggested to indicate infection of postoperative abdominal fluid collections; however
Cécile R L P N Jeukens et al.
Investigative radiology, 49(10), 659-665 (2014-05-30)
Contrast-enhanced spectral mammography (CESM) shows promising initial results but comes at the cost of increased dose as compared with full-field digital mammography (FFDM). We aimed to quantitatively assess the dose increase of CESM in comparison with FFDM. Radiation exposure-related data
J Kooiman et al.
Journal of thrombosis and haemostasis : JTH, 12(10), 1658-1666 (2014-08-22)
Hydration to prevent contrast-induced acute kidney injury (CI-AKI) induces a diagnostic delay when performing computed tomography-pulmonary angiography (CTPA) in patients suspected of having acute pulmonary embolism. To analyze whether withholding hydration is non-inferior to sodium bicarbonate hydration before CTPA in
Nicholas M Orme et al.
Mayo Clinic proceedings, 89(6), 747-753 (2014-06-20)
To determine the frequency and clinical impact of noncardiovascular incidental findings (IFs) detected on preoperative computed tomographic angiography (CTA) of the chest/abdomen/pelvis performed in elderly patients with severe aortic stenosis being considered for transcatheter aortic valve replacement (TAVR). The CTA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service