Skip to Content
Merck
All Photos(1)

Key Documents

Y0001021

Iopromide for system suitability 1

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Iopromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24I3N3O8
CAS Number:
Molecular Weight:
791.11
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

iopromide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C(N(C)CC(O)CO)C1=C(I)C(NC(COC)=O)=C(I)C(C(NCC(CO)O)=O)=C1I

InChI

1S/C18H24I3N3O8/c1-24(4-9(28)6-26)18(31)12-13(19)11(17(30)22-3-8(27)5-25)14(20)16(15(12)21)23-10(29)7-32-2/h8-9,25-28H,3-7H2,1-2H3,(H,22,30)(H,23,29)

InChI key

DGAIEPBNLOQYER-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Iopromide for system suitability 1 EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cécile R L P N Jeukens et al.
Investigative radiology, 49(10), 659-665 (2014-05-30)
Contrast-enhanced spectral mammography (CESM) shows promising initial results but comes at the cost of increased dose as compared with full-field digital mammography (FFDM). We aimed to quantitatively assess the dose increase of CESM in comparison with FFDM. Radiation exposure-related data
Nicholas M Orme et al.
Mayo Clinic proceedings, 89(6), 747-753 (2014-06-20)
To determine the frequency and clinical impact of noncardiovascular incidental findings (IFs) detected on preoperative computed tomographic angiography (CTA) of the chest/abdomen/pelvis performed in elderly patients with severe aortic stenosis being considered for transcatheter aortic valve replacement (TAVR). The CTA
J Kooiman et al.
Journal of thrombosis and haemostasis : JTH, 12(10), 1658-1666 (2014-08-22)
Hydration to prevent contrast-induced acute kidney injury (CI-AKI) induces a diagnostic delay when performing computed tomography-pulmonary angiography (CTPA) in patients suspected of having acute pulmonary embolism. To analyze whether withholding hydration is non-inferior to sodium bicarbonate hydration before CTPA in
Felix G Meinel et al.
Radiology, 270(3), 708-716 (2014-01-31)
To compare the relative contributions of rest, stress, and delayed acquisitions with the accuracy of dual-energy (DE) computed tomography (CT) for the assessment of myocardial blood supply. With institutional review board approval and HIPAA compliance, 55 consecutive patients (10 women
Michel Habis et al.
Radiology, 274(3), 684-692 (2015-01-20)
To assess the feasibility of exercise perfusion computed tomography (CT) in patients suspected of having hemodynamically significant coronary stenosis. This study had institutional review board approval, and all patients gave informed consent. Thirty-two consecutive patients (26 men [mean age, 63

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service